Skip to Content
Merck
All Photos(1)

Documents

431877

Sigma-Aldrich

Methyl 5-methylsalicylate

96%

Synonym(s):

Methyl 2-hydroxy-5-methylbenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(OH)CO2CH3
CAS Number:
Molecular Weight:
166.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

96%

refractive index

n20/D 1.533 (lit.)

bp

241-243 °C/767 mmHg (lit.)

density

1.153 g/mL at 25 °C (lit.)

SMILES string

COC(=O)c1cc(C)ccc1O

InChI

1S/C9H10O3/c1-6-3-4-8(10)7(5-6)9(11)12-2/h3-5,10H,1-2H3

InChI key

JQYUQKRFSSSGJM-UHFFFAOYSA-N

General description

Methyl 5-methylsalicylate is an ester. It acts as ligand and forms novel nonanuclear Tb(III) clusters. The photophysical properties of these clusters have been studied.

Application

Methyl 5-methylsalicylate is the suitable reagent employed in study for the identification of a pheromone component in parasitoid wasp Spalangia endius. It may be used for the synthesis of 2,6-dimethylchromanone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

226.4 °F - closed cup

Flash Point(C)

108 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Conjugate addition to 3-arylsulfinylchromones as a synthetic route to homochiral 2-substituted chromanones: scope and limitations.
Hodgetts KJ, et al.
Tetrahedron, 57(31), 6793-6804 (2001)
Shun Omagari et al.
The journal of physical chemistry. A, 119(10), 1943-1947 (2015-02-12)
The photophysical properties of the novel nonanuclear Tb(III) clusters Tb-L1 and Tb-L2 involving the ligands methyl 4-methylsalicylate (L1) and methyl 5-methylsalicylate (L2) are reported. The position of the methyl group has an effect on their photophysical properties. The prepared nonanuclear
William J Nichols et al.
Journal of chemical ecology, 36(10), 1140-1147 (2010-09-08)
Sex-pheromone-related behavior and chemistry were studied in the wasp Spalangia endius Walker (Hymenoptera: Pteromalidae), a pupal parasitoid of the house fly, Musca domestica L. (Diptera: Muscidae). Males responded behaviorally to female extracts by arrestment, whereas females did not arrest to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service