Skip to Content
Merck
All Photos(1)

Key Documents

431044

Sigma-Aldrich

(Heptafluoropropyl)trimethylsilane

97%

Synonym(s):

1-(Trimethylsilyl)heptafluoropropane

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€118.00

€118.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€118.00

About This Item

Linear Formula:
CF3CF2CF2Si(CH3)3
CAS Number:
Molecular Weight:
242.21
Beilstein:
1769415
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

€118.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.324 (lit.)

bp

88 °C (lit.)

density

1.195 g/mL at 25 °C (lit.)

functional group

fluoro

SMILES string

C[Si](C)(C)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C6H9F7Si/c1-14(2,3)6(12,13)4(7,8)5(9,10)11/h1-3H3

InChI key

PKLASFRWKXWKII-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in:
  • Trifluoromethylation of arenes
  • Nucleophilic addition followed by intramolecular aza-Michael reactions
  • Perfluoroalkylation of carbonyl compounds, aldimines and other organic substrates
Used to prepare perfluoroalkyl compounds[1] and heptafluoro-n-propyl derivatives of phosphorus and silicon.[2]

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. Ind. Chem. Soc., 55, 1254-1254 (1978)
The Journal of Organic Chemistry, 56, 984-984 (1991)
Eleonora Perego et al.
Lab on a chip, 21(4), 735-745 (2021-01-26)
Despite the importance for cellular processes, the dynamics of molecular assembly, especially on fast time scales, is not yet fully understood. To this end, we present a multi-layer microfluidic device and combine it with fluorescence fluctuation spectroscopy. We apply this

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service