Skip to Content
Merck
All Photos(1)

Documents

430986

Sigma-Aldrich

1,2-Dihydronaphthalene

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10
CAS Number:
Molecular Weight:
130.19
Beilstein:
1851372
EC Number:
MDL number:
UNSPSC Code:
12352100

Assay

≥99%

refractive index

n20/D 1.582 (lit.)

bp

89 °C/16 mmHg (lit.)

mp

−8 °C (lit.)

density

0.997 g/mL at 25 °C (lit.)

SMILES string

C1Cc2ccccc2C=C1

InChI

1S/C10H10/c1-2-6-10-8-4-3-7-9(10)5-1/h1-3,5-7H,4,8H2

InChI key

KEIFWROAQVVDBN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

152.6 °F - closed cup

Flash Point(C)

67 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D S Torok et al.
Journal of bacteriology, 177(20), 5799-5805 (1995-10-01)
Bacterial strains expressing toluene and naphthalene dioxygenase were used to examine the sequence of reactions involved in the oxidation of 1,2-dihydronaphthalene. Toluene dioxygenase of Pseudomonas putida F39/D oxidizes 1,2-dihydronaphthalene to (+)-cis-(1S,2R)-dihydroxy-1,2,3,4-tetrahydronaphthalene, (+)-(1R)-hydroxy-1,2-dihydronaphthalene, and (+)-cis-(1R,2S)-dihydroxy-1,2-dihydronaphthalene. In contrast, naphthalene dioxygenase of Pseudomonas
Keith Smith et al.
Chemical communications (Cambridge, England), (8)(8), 886-887 (2002-07-19)
We have successfully prepared an unsymmetrical analogue of a Katsuki-type salen ligand having a single hydroxyalkyl group at its 6-position, and also its Mn(III) complex; attachment of the complex to a polymer gives a highly enantioselective and recoverable catalyst for
S L Eaton et al.
Applied and environmental microbiology, 62(12), 4388-4394 (1996-12-01)
The substrate oxidation profiles of Sphingomonas yanoikuyae B1 biphenyl-2,3-dioxygenase and cis-biphenyl dihydrodiol dehydrogenase activities were examined with 1,2-dihydronaphthalene and various cis-diols as substrates. m-Xylene-induced cells of strain B1 oxidized 1,2-dihydronaphthalene to (-)-(1R,2S)-cis-1,2-dihydroxy-1,2-3,4-tetrahydronaphthalene as the major product (73% relative yield). Small
Claudia Sanfilippo et al.
Biotechnology letters, 26(23), 1815-1819 (2005-01-27)
Chloroperoxidase from Caldariomyces fumago catalyses the oxidation of 1,2-dihydronaphthalene to (1R,2R)-(+)-dihydroxytetrahydronaphthalene in homogenous citrate buffer/ionic liquid mixtures, using t-butyl hydroperoxide as O2 donor. It tolerates up to 30 (v/v) 1,3-dimethylimidazolium methylsulfate or 1-butyl-3-methylimidazolium methylsulfate. The enzyme activity in these ionic
Márcia Kameyama et al.
Molecules (Basel, Switzerland), 16(11), 9421-9438 (2011-11-15)
A new approach for the synthesis of indatraline was developed using as the key step an iodine(III)-mediated ring contraction of a 1,2-dihydronaphthalene derivative. Behavioral tests were conducted to evaluate the effect of indatraline and of its precursor indanamide on the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service