Skip to Content
Merck
All Photos(1)

Key Documents

42995

Sigma-Aldrich

trans-1,3-Diphenyl-2,3-epoxypropan-1-one

≥98.0%

Synonym(s):

trans-2-Benzoyl-3-phenyloxirane, trans-Benzalacetophenone oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12O2
CAS Number:
Molecular Weight:
224.25
Beilstein:
84132
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0%

form

solid

mp

86-89 °C (lit.)

functional group

ether
ketone
phenyl

SMILES string

O=C([C@H]1O[C@@H]1c2ccccc2)c3ccccc3

InChI

1S/C15H12O2/c16-13(11-7-3-1-4-8-11)15-14(17-15)12-9-5-2-6-10-12/h1-10,14-15H/t14-,15-/m1/s1

InChI key

UQGMJZQVDNZRKT-HUUCEWRRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

trans-1,3-Diphenyl-2,3-epoxypropan-1-one has been reported to efficiently participate in gas-phase Meerwein reaction under both electrospray ionization (ESI) and atmospheric pressure chemical ionization (APCI) conditions. Therapeutic potential of trans-1,3-diphenyl-2,3-epoxypropane-1-one (DPEP), and its anti-inflammatory effects have been studied.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Seidegård et al.
European journal of biochemistry, 112(3), 643-648 (1980-12-01)
Benzil was found to be a very potent activator of microsomal epoxide hydrolase activity (measured with styrene oxide as substrate) in vitro. The activating effect was uncompetitive and benzil causes approximately ninefold increases in both the apparent V and the
G D Prestwich et al.
Archives of biochemistry and biophysics, 242(1), 11-15 (1985-10-01)
Chalcone oxides and several isosteric compounds have been prepared to examine the importance of the alpha,beta-epoxyketone moiety in the inhibition of the hydrolysis of [3H]-trans-stilbene oxide to its meso-diol by mouse liver cytosolic epoxide hydrolase (cEH). Inhibition of microsomal EH
Giulia Licini et al.
Biopolymers, 84(1), 97-104 (2005-08-27)
C(alpha)-tetrasubstituted alpha-amino acids constitute a powerful tool for controlling the conformation of short peptide sequences. Chiral peptides may be used in stereoselective reactions both for asymmetric induction and in kinetic resolution. By reviewing recent data from our own laboratories and
E C Dietze et al.
Biochemical pharmacology, 42(6), 1163-1175 (1991-08-22)
The inhibition of murine cytosolic epoxide hydrolase has been studied with both racemic and enantiomerically pure trans-3-phenylglycidols. These compounds are the first enantioselective, slow binding inhibitors of cytosolic epoxide hydrolase. The (2S,3S)-3-phenylglycidol enantiomer was always a better inhibitor than the
Chalcone oxides--potent selective inhibitors of cytosolic epoxide hydrolase.
C A Mullin et al.
Archives of biochemistry and biophysics, 216(2), 423-439 (1982-07-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service