42830
2,3-Diphenyl-2,3-butanediol
technical, mixture of racemic and meso forms, ≥90% (GC)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6H5C(CH3)(OH)C(OH)(CH3)C6H5
CAS Number:
Molecular Weight:
242.31
Beilstein:
1878154
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
grade
technical
Assay
≥90% (GC)
SMILES string
CC(O)(c1ccccc1)C(C)(O)c2ccccc2
InChI
1S/C16H18O2/c1-15(17,13-9-5-3-6-10-13)16(2,18)14-11-7-4-8-12-14/h3-12,17-18H,1-2H3
InChI key
URPRLFISKOCZHR-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
2,3-Diphenyl-2,3-butanediol is a pinacol. A mixture of meso and rac-2,3-diphenyl-2,3-butanediol is formed during the reduction of acetophenone. 2,3-Diphenyl-2,3-butanediol forms 2,3-diphenyl-1,3-butadiene by dehydration.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Mechanism of electrochemical pinacolization. The first asymmetric synthesis in a chiral medium.
Seebach D and Oei HA.
Angewandte Chemie (International Edition in English), 14(9), 634-636 (1975)
Dialkyl Peroxide-Induced Reductions of Aromatic Ketones.
Huyser ES and Neckers DC.
Journal of the American Chemical Society, 85(22), 3641-3644 (1963)
Head to head polymers-38. An improved synthesis of head to head polystyrene.
Foldes E, et al.
European Polymer Journal, 29(2), 321-330 (1993)
Comparative study of the NMR length scale of dynamic heterogeneities of three different glass formers.
Reinsberg SA, et al.
Journal of Non-Crystalline Solids, 307, 208-214 (2002)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service