Skip to Content
Merck
All Photos(3)

Key Documents

425370

Sigma-Aldrich

Phenol-2,3,4,5,6-d5

98 atom % D

Synonym(s):

Pentadeuteriophenol

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€216.00
5 G
€735.00

€216.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 G
€216.00
5 G
€735.00

About This Item

Linear Formula:
C6D5OH
CAS Number:
Molecular Weight:
99.14
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.12

€216.00


Check Cart for Availability

Request more information

isotopic purity

98 atom % D

Quality Level

form

crystals

bp

182 °C (lit.)

mp

40-42 °C (lit.)

density

1.127 g/mL at 25 °C

mass shift

M+5

SMILES string

[2H]c1c([2H])c([2H])c(O)c([2H])c1[2H]

InChI

1S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7H/i1D,2D,3D,4D,5D

InChI key

ISWSIDIOOBJBQZ-RALIUCGRSA-N

General description

Phenol-2,3,4,5,6-d5 is an isotope-labeled analog of phenol, wherein 2,3,4, 5, and 6th protons of phenol are replaced by deuterium.

Application

Phenol-2,3,4,5,6-d5 can be used as an internal analytical standard for accurate data interpretation using various analytical techniques.[1][2]

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - STOT RE 2

Target Organs

Nervous system,Kidney,Liver,Skin

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Nature of meso-and pyro-Borate Precatalysts to the VANOL and VAPOL BOROX Catalysts
Zhao W, et al.
Synlett, 26, 1606-1614 (2015)
Development of molecularly imprinted polymer in porous film format for binding of phenol and alkylphenols from water
Gryshchenko AO and Bottaro CS
International Journal of Molecular Sciences, 15, 1338-1357 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service