Skip to Content
Merck
All Photos(2)

Key Documents

424927

Sigma-Aldrich

Propylene glycol propyl ether

99%

Synonym(s):

1-Propoxy-2-propanol

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€54.80
4 L
€143.00
18 L
€400.00

€54.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 L
€54.80
4 L
€143.00
18 L
€400.00

About This Item

Linear Formula:
CH3CH2CH2OCH2CH(OH)CH3
CAS Number:
Molecular Weight:
118.17
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€54.80


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.411 (lit.)

bp

140-160 °C (lit.)

density

0.885 g/mL at 25 °C (lit.)

SMILES string

CCCOCC(C)O

InChI

1S/C6H14O2/c1-3-4-8-5-6(2)7/h6-7H,3-5H2,1-2H3

InChI key

FENFUOGYJVOCRY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

115.5 °F - closed cup

Flash Point(C)

46.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Ballantyne et al.
Veterinary and human toxicology, 30(2), 126-129 (1988-04-01)
1-Propoxy-2-propanol, a widely used industrial chemical, was found to have acute peroral LD50 values in the rat of 4.92 ml/kg (males) and 2.83 ml/kg (females), with the signs of systemic toxicity being principally related to narcosis. Acute percutaneous LD50 values
Yanqin Gao et al.
The Journal of chemical physics, 142(21), 214505-214505 (2015-06-08)
The dielectric relaxations in six primary and secondary alkoxy alcohols with varying molecular size and different separation between -O- and hydroxyl group are studied at temperatures around glass transition. The analyses of the apparent full width at half maximum of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service