Skip to Content
Merck
All Photos(1)

Key Documents

424773

Sigma-Aldrich

4-Benzylidene-2-phenyl-2-oxazolin-5-one

≥99%

Synonym(s):

(Z)-4-Benzylidene-2-phenyloxazolin-5-one, (Z)-4-benzylidene-2-phenyloxazol-5(4H)-one, 4-Benzylidene-2-phenyl-5(4H)-oxazolone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H11NO2
CAS Number:
Molecular Weight:
249.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥99%

mp

165-167 °C (lit.)

SMILES string

O=C1OC(=N\C1=C/c2ccccc2)c3ccccc3

InChI

1S/C16H11NO2/c18-16-14(11-12-7-3-1-4-8-12)17-15(19-16)13-9-5-2-6-10-13/h1-11H/b14-11-

InChI key

VFDOKJVMHZUBTN-KAMYIIQDSA-N

General description

4-Benzylidene-2-phenyl-2-oxazolin-5-one (4-benzal-2-phenyl-5-oxazolone) is an azlactone. The absolute configuration of the geometric isomers of 4-benzylidene-2-phenyl-2-oxazolin-5-one have been determined by NMR spectroscopy.[1] It undergoes two primary photochemical processes, geometric isomerization and hydrogen abstraction from solvent.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Unsaturated lactone photochemistry. Effect of wavelength and sensitizer structure on selective population of specific excited states.
Ullman EF and Baumann N.
Journal of the American Chemical Society, 92(20), 5892-5899 (1970)
The geometrical isomers of 4-benzylidene-2-phenyl-2-oxazolin-5-one: determination of absolute configurations by nuclear magnetic resonance spectroscopy.
Brocklehurst K, et al.
Chemical Communications (Cambridge, England), 15, 884-886 (1968)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service