Skip to Content
Merck
All Photos(1)

Key Documents

423807

Sigma-Aldrich

5-Hydroxy-2-adamantanone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14O2
CAS Number:
Molecular Weight:
166.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

>300 °C (lit.)

functional group

hydroxyl
ketone

SMILES string

O[C@]12C[C@@H]3C[C@H](C1)C(=O)[C@@H](C3)C2

InChI

1S/C10H14O2/c11-9-7-1-6-2-8(9)5-10(12,3-6)4-7/h6-8,12H,1-5H2/t6-,7-,8+,10-

InChI key

TZBDEVBNMSLVKT-XYYXLIQBSA-N

General description

5-Hydroxy-2-adamantanone is a disubstituted derivative of adamantane.[1] The biocatalyzed synthesis of 5-hydroxy-2-adamantanone from 2-adamantanone has been investigated.[2]

Application

5-Hydroxy-2-adamantanone may be used in the following studies:
  • As a model compound to investigate the application of lanthanide NMR shift reagents for the analysis of disubstituted derivative of adamantane.[1]
  • As a starting material for the synthesis of E-2-amino-5-hydroxyadamantane.[3]
  • As a starting material for the synthesis of 4-(triphenylsilyloxy)adamantan-1-ol.[4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[The immunomodulator kemantan in the treatment of patients with exacerbated chronic obstructive bronchitis].
E M Rekalova
Likars'ka sprava, (4)(4), 73-76 (1992-04-01)
S S Boĭko et al.
Farmakologiia i toksikologiia, 54(1), 57-59 (1991-01-01)
The pharmacokinetics of a new Soviet-made immunostimulant kemantane, a derivative of adamantine, was studied by gas-liquid chromatography in patients with bronchial pathology. It was found that in the blood of the patients kemantane was not practically detected due to a
An expeditious preparation of E-2-amino-5-hydroxyadamantane and its Z-isomer.
Jaroskova L, et al.
Tetrahedron Letters, 47(46), 8063-8067 (2006)
Marta L Lage et al.
Tetrahedron, 69(27-28), 5609-5613 (2013-09-03)
A chemoselective method for the hydrosilylation of ketones has been developed, using the combination of triphenylsilane and a catalyst prepared from Ni(COD)
Application of shift reagents in the study of disubstituted derivatives of adamantane by NMR spectroscopy.
Vodicka L, et al.
Collection of Czechoslovak Chemical Communications, 40(1), 293-299 (1975)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service