Skip to Content
Merck
All Photos(2)

Key Documents

423726

Sigma-Aldrich

N-Z-L-aspartic anhydride

95%

Synonym(s):

Cbz-L-Aspartic acid anhydride, N-(Benzyloxycarbonyl)-L-aspartic acid anhydride, Phenylmethyl [(3S)-tetrahydro-2,5-dioxo-3-furanyl]carbamate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€204.00

€204.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€204.00

About This Item

Empirical Formula (Hill Notation):
C12H11NO5
CAS Number:
Molecular Weight:
249.22
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

€204.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

powder

optical activity

[α]20/D −25°, c = 1 in methanol

reaction suitability

reaction type: solution phase peptide synthesis

mp

123-124 °C (lit.)

application(s)

peptide synthesis

SMILES string

O=C1C[C@H](NC(=O)OCc2ccccc2)C(=O)O1

InChI

1S/C12H11NO5/c14-10-6-9(11(15)18-10)13-12(16)17-7-8-4-2-1-3-5-8/h1-5,9H,6-7H2,(H,13,16)/t9-/m0/s1

InChI key

OZPYEGOBGWQOSZ-VIFPVBQESA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nadia B Haro-Mares et al.
Molecules (Basel, Switzerland), 25(13) (2020-07-09)
A simplified procedure to synthesize zwitterionic cellulose by means of N-protected aspartic anhydride under mild conditions was developed. The preparation of modified cellulose samples was carried out under heterogeneous, aqueous conditions by reacting NH4OH-activated cellulose with aspartic anhydrides N-protected with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service