Skip to Content
Merck
All Photos(1)

Key Documents

423556

Sigma-Aldrich

1,4,7,10-Tetratosyl-1,4,7,10-tetraazacyclododecane

technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H44N4O8S4
CAS Number:
Molecular Weight:
789.02
Beilstein:
603592
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

grade

technical grade

mp

275-279 °C (lit.)

SMILES string

Cc1ccc(cc1)S(=O)(=O)N2CCN(CCN(CCN(CC2)S(=O)(=O)c3ccc(C)cc3)S(=O)(=O)c4ccc(C)cc4)S(=O)(=O)c5ccc(C)cc5

InChI

1S/C36H44N4O8S4/c1-29-5-13-33(14-6-29)49(41,42)37-21-23-38(50(43,44)34-15-7-30(2)8-16-34)25-27-40(52(47,48)36-19-11-32(4)12-20-36)28-26-39(24-22-37)51(45,46)35-17-9-31(3)10-18-35/h5-20H,21-28H2,1-4H3

InChI key

SKTWVKMQYXXWNY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service