Skip to Content
Merck
All Photos(1)

Key Documents

423343

Sigma-Aldrich

p-Tolyl [2-(trimethylsilyl)ethynyl] sulfone

98%

Synonym(s):

1-(p-Toluenesulfonyl)-2-(trimethylsilyl)acetylene, 2-(Trimethylsilyl)ethynyl p-tolyl sulfone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiC≡CSO2C6H4CH3
CAS Number:
Molecular Weight:
252.40
Beilstein:
2650368
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
98%

Assay

98%

mp

80-83 °C (lit.)

storage temp.

2-8°C

SMILES string

Cc1ccc(cc1)S(=O)(=O)C#C[Si](C)(C)C

InChI

1S/C12H16O2SSi/c1-11-5-7-12(8-6-11)15(13,14)9-10-16(2,3)4/h5-8H,1-4H3

InChI key

MMUNNGUGQXEUBR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

p-Tolyl [2-(trimethylsilyl)ethynyl] sulfone is an aryl trimethylsilylethynyl sulfone. It can be synthesized from bis(trimethylsilyl)acetylene.[1]

Application

p-Tolyl [2-(trimethylsilyl)ethynyl] sulfone (1-tosyl-2-(trimethylsilyl)acetylene) may be used as a reactant in the synthesis of Cp(OC)2M-C=CSiMe3 (M = Fe, Ru) by reacting with [Cp(OC)2M ]-Na+[2] and as a reagent in the photochemical alkynylation of unreactive C (sp3)–H bonds.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photochemically induced radical alkynylation of C (sp3)-H bonds.
Hoshikawa T, et al.
Organic & Biomolecular Chemistry, 11(1), 164-169 (2012)
p-Tolyl(trimethylsiIylethynyl)sulfone as Synthon for the Synthesis of Cp(OC)2M -C=CSiMe3 (M = Fe, Ru). Molecular Structure of Cp(OC)2Fe-C=CSiMe3
Weinrich V and Beck W.
Zeitschrift fur Naturforschung, 54, 701-703 (1999)
King RB.
Organometallic Syntheses, 4, 466-466 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service