Skip to Content
Merck
All Photos(1)

Key Documents

422754

Sigma-Aldrich

3-Decyn-1-ol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5C≡CCH2CH2OH
CAS Number:
Molecular Weight:
154.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.459 (lit.)

bp

130-131 °C/22 mmHg (lit.)

density

0.877 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CCCCCCC#CCCO

InChI

1S/C10H18O/c1-2-3-4-5-6-7-8-9-10-11/h11H,2-6,9-10H2,1H3

InChI key

YGEQBZUDPQQIFI-UHFFFAOYSA-N

General description

3-Decyn-1-ol is primary homo-propargylic alcohol derivative. It has been reported to be formed during the isomerization of 2-decyn-1-ol in the presence of sodium salt of 1,3-diaminopropane. Its toxicity has been evaluated on the green alga, Pseudokirchneriella subcapitata in terms of EC50 (median effective concentration) and NOEC (no observed effect level) values. It undergoes semi hydrogenation in the presence of Lindlar catalyst to form cis-3-decen-1-ol.

Application

3-Decyn-1-ol may be used in the following studies:
  • As a starting material in the synthesis of 3′-deoxyribolactones.
  • As a starting material in the synthesis of (Z)-3-decen-1-ol by hydrogenation over P-2 nickel.
  • As a reagent in the synthesis of methyl 9-decynoate.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

224.6 °F - closed cup

Flash Point(C)

107 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 3'-Deoxyribolactones using a Hydrolysis-Induced Lactonization Cascade Reaction of Epoxy Cyanohydrins.
Vugts DJ, et al.
European Journal of Organic Chemistry, 2008(8), 1336-1339 (2008)
Miriam F Cooperband et al.
Journal of chemical ecology, 38(1), 52-62 (2012-01-17)
A male-produced pheromone that attracts both males and females was identified for the European woodwasp, Sirex noctilio, a serious pest of pine trees. Males displayed excitatory behaviors when placed in groups, and were attracted to the odors from males that
The rearrangement of isomeric linear decyn-1-ols by reaction with the sodium salt of 1, 3-diaminopropane.
Macaulay SR.
Canadian Journal of Chemistry, 58(23), 2567-2572 (1980)
Seiji Itoh et al.
Bioscience, biotechnology, and biochemistry, 66(7), 1591-1596 (2002-09-13)
Both the (17R)- and (17S)-isomers of volicitin, which is contained in the oral secretion of the beet armyworm and induces corn seedlings to emit a blend of volatile compounds to attract the natural enemy of the herbivore, were synthesized via
Chung Yuan Chen et al.
Journal of environmental monitoring : JEM, 14(1), 181-186 (2011-11-23)
The present study evaluates the toxicity of 34 propargylic alcohols, including primary, primary homo-, secondary, and tertiary alcohols, based on their effects on phytoplankton. A closed-system algal toxicity test was applied because the closed-system technique presents more realistic concentration-response relationships

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service