Skip to Content
Merck
All Photos(1)

Documents

422436

Sigma-Aldrich

Methyl 1-cyclopentene-1-carboxylate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H7CO2CH3
CAS Number:
Molecular Weight:
126.15
Beilstein:
2040162
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.4660 (lit.)

bp

76-78 °C/9 mmHg (lit.)

density

1.031 g/mL at 20 °C (lit.)

SMILES string

COC(=O)C1=CCCC1

InChI

1S/C7H10O2/c1-9-7(8)6-4-2-3-5-6/h4H,2-3,5H2,1H3

InChI key

VTYCAXIAUKEGBQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl 1-cyclopentene-1-carboxylate, a cyclic alkene, is a cyclopentene derivative. It participates in the synthesis of azulene derivatives by initially forming the corresponding cyclopropanol followed by oxy-Cope rearrangement.

Application

Methyl 1-cyclopentene-1-carboxylate may be used as starting material in the synthesis of pinnaic acid and halichlorine. It undergoes asymmetric oxidative Heck reaction with aryl boronic acids to form coupling products in the presence of chiral NHC (N-heterocyclic carbine)-palladium (II) complex.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

THE ORGANIC CHEMISTRY NOTEBOOK SERIES, A DIDACTICAL APPROACH,?(I) A THEORETICAL MECHANISTIC APPROACH TO DIASTEROSELECTIVE SYNTHESIS OF CIS-1, 2-DIALKENYLCYCLOPROPANOLS AND SUBSEQUENT OXY-COPE REARRANGEMENT BY JIN KUN CHA ET AL.
Bravo J.
Revista Boliviana de Quimica, 22(1), 1-10 (2005)
Rodrigo B Andrade et al.
Organic letters, 7(25), 5733-5735 (2005-12-03)
[chemical reaction: see text]. Concise formal syntheses of marine alkaloids (+/-)-pinnaic acid (1) and (+/-)-halichlorine (2) have been accomplished from a common intermediate. The syntheses illustrate the utility of selective olefin cross metathesis methodologies for the elaboration of advanced synthetic
Kyung Soo Yoo et al.
The Journal of organic chemistry, 75(1), 95-101 (2009-12-04)
Chiral dimeric tridentate NHC-amidate-alkoxide palladium(II) complexes, 3a and 3b, effected oxidative boron Heck-type reactions of aryl boronic acids with both acyclic and cyclic alkenes at room temperature to afford the corresponding coupling products with high enantioselectivities. The high degree of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service