Skip to Content
Merck
All Photos(1)

Key Documents

42155

Sigma-Aldrich

2,3-Dinitrophenol

moistened with water, ≥98.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4N2O5
CAS Number:
Molecular Weight:
184.11
Beilstein:
2213580
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (HPLC)

quality

moistened with water

contains

~20% water as stabilizer

mp

141-145 °C

SMILES string

Oc1cccc(c1[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C6H4N2O5/c9-5-3-1-2-4(7(10)11)6(5)8(12)13/h1-3,9H

InChI key

MHKBMNACOMRIAW-UHFFFAOYSA-N

General description

2,3-Dinitrophenol is a nitroaromatic compound.[1] It has been reported to exhibit high acute toxicity to Scenedesmus obliquus, with EC50 value of 2.5±0.8.[2]

Application

2,3-Dinitrophenol may be employed as nitrogen supplement for the bacterial culture medium.[1]

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Expl. 1.1 - STOT RE 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marcus M Fischer et al.
Scientific reports, 7(1), 15270-15270 (2017-11-12)
Activating mutations in the Wnt pathway are a characteristic feature of colorectal cancer (CRC). The R-spondin (RSPO) family is a group of secreted proteins that enhance Wnt signaling and RSPO2 and RSPO3 gene fusions have been reported in CRC. We
David Schmidt et al.
PLoS genetics, 14(12), e1007828-e1007828 (2018-12-18)
In multicellular organisms, cell type diversity and fate depend on specific sets of transcript isoforms generated by post-transcriptional RNA processing. Here, we used Schmidtea mediterranea, a flatworm with extraordinary regenerative abilities and a large pool of adult stem cells, as
C Bruhn et al.
Applied and environmental microbiology, 53(1), 208-210 (1987-01-01)
Bacteria which utilized nitroaromatic compounds (0.5 mM) as sole source of nitrogen were isolated from soil. With 2,6-dinitrophenol and succinate as carbon source, a Pseudomonas strain was isolated which liberated and assimilated nitrite. Approximately 2 mol of NO(2) per mol
María Almuedo-Castillo et al.
Nature cell biology, 20(9), 1032-1042 (2018-08-01)
Individuals can vary substantially in size, but the proportions of their body plans are often maintained. We generated smaller zebrafish by removing 30% of their cells at the blastula stages and found that these embryos developed into normally patterned individuals.
Katharina Boroviak et al.
Scientific reports, 7(1), 12867-12867 (2017-10-11)
Modelling human diseases caused by large genomic rearrangements has become more accessible since the utilization of CRISPR/Cas9 in mammalian systems. In a previous study, we showed that genomic rearrangements of up to one million base pairs can be generated by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service