Skip to Content
Merck
All Photos(2)

Key Documents

418706

Sigma-Aldrich

5-Ethyl-1,3-dioxane-5-methanol

98%

Synonym(s):

(5-Ethyl[1,3]dioxan-5-yl)methanol, 5-(Hydroxymethyl)-5-ethyl-1,3-dioxane, 5-Ethyl-1,3-dioxan-5-methanol, 5-Ethyl-1,3-dioxane-5-methanol, 5-Ethyl-5-(hydroxymethyl)-1,3-dioxane, 5-Ethyl-m-dioxane-5-methanol, Formaldehyde mono(1,1,1-trimethylolpropane) acetal

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
€126.00

€126.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 ML
€126.00

About This Item

Empirical Formula (Hill Notation):
C7H14O3
CAS Number:
Molecular Weight:
146.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€126.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

refractive index

n20/D 1.463 (lit.)

bp

104-105 °C/5 mmHg (lit.)

density

1.09 g/mL at 25 °C (lit.)

functional group

ether
hydroxyl

SMILES string

CCC1(CO)COCOC1

InChI

1S/C7H14O3/c1-2-7(3-8)4-9-6-10-5-7/h8H,2-6H2,1H3

InChI key

BGFBWRWYROQISE-UHFFFAOYSA-N

General description

Physical properties (such as boiling point, refractive index and density) of 5-ethyl-1,3-dioxane-5-methanol have been reported.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

240.8 °F - closed cup

Flash Point(C)

116 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaws CL
The Yaws Handbook of Physical Properties for Hydrocarbons and Chemicals, 169-169 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service