Skip to Content
Merck
All Photos(2)

Key Documents

418005

Sigma-Aldrich

Poly(hexamethylene diisocyanate)

viscosity 1,300-3,000 cP (25 °C)(lit.)

Synonym(s):

1,6-Diisocyanatohexane homopolymer, 1,6-Diisocyanatohexane polymer, HMDI homopolymer

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€72.20

€72.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€72.20

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€72.20


Check Cart for Availability

Request a Bulk Order

form

liquid

Quality Level

refractive index

n20/D 1.502

viscosity

1,300-3,000 cP(25 °C)(lit.)

density

1.12 g/mL at 25 °C

SMILES string

O=C=NCCCCCCN=C=O

InChI key

RRAMGCGOFNQTLD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(hexamethylene diisocyanate) (PolyHDI) is an aliphatic compound belonging to the class of isocyanates. It is generally prepared by reacting hexamethylene diamine with phosgene.[1]

Application

PolyHDI can be used as a compatibilizer to increase the complex viscosity of poly(3-hydroxybutyrate-co-3-hydroxyvalerate) (PHBV)/poly(lactic acid) biopolymer based blends.[1] It can be used as a hardener that can be mixed with polyol resins to form polyurethane coatings with a wide range of applications like self-cleaning.[2][3][4]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

338.0 °F

Flash Point(C)

170 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antimicrobial polyurethane foams having cationic ammonium groups
Udabe E, et al.
Journal of Applied Polymer Science, 134(45), 45473-45473 (2017)
Compatibilization of poly (3-hydroxybutyrate-co-3-hydroxyvalerate)-poly (lactic acid) blends with diisocyanates
Gonzalez-Ausejo J, et al.
Journal of Applied Polymer Science, 134(20) (2017)
Structured polyurethanes for oil uptake
Yang JS, et al.
Journal of Applied Polymer Science, 98(5), 2080-2087 (2005)
SiO2 encapsulated TiO2 nanotubes and nanofibers for self-cleaning polyurethane coatings
Chen C, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 348(5), 226-237 (2017)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service