Skip to Content
Merck
All Photos(1)

Key Documents

417793

Sigma-Aldrich

5-Ethyl-1-aza-3,7-dioxabicyclo[3.3.0]octane

97%

Synonym(s):

5-Ethyl-3,7-dioxa-1-azabicyclo[3.3.0]octane, Dihydro-7a-ethyloxazolo[3,4-c]oxazole

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€68.50

€68.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
€68.50

About This Item

Empirical Formula (Hill Notation):
C7H13NO2
CAS Number:
Molecular Weight:
143.18
Beilstein:
105946
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€68.50


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

liquid

refractive index

n20/D 1.461 (lit.)

bp

75 °C/10 mmHg (lit.)

density

1.083 g/mL at 25 °C (lit.)

SMILES string

CCC12COCN1COC2

InChI

1S/C7H13NO2/c1-2-7-3-9-5-8(7)6-10-4-7/h2-6H2,1H3

InChI key

ZRCMGIXRGFOXNT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • A new formulation for the treatment of acid-deterioration (Red Rot) in historic leathers: This study introduces a new formulation containing 5-ethyl-1-aza-3,7-dioxabicyclo[3.3.0]octane for treating acid-deteriorated historic leather, highlighting its potential in preserving cultural heritage materials (A Lama et al., 2015).
  • Use of aluminium alkoxide and oxazolidine II to treat acid-deteriorated historic leather: Examines the effectiveness of a treatment combining 5-ethyl-1-aza-3,7-dioxabicyclo[3.3.0]octane with aluminium alkoxide on historic leather, comparing it to individual treatments to assess synergistic effects (A Lama et al., 2015).

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natasha Babar et al.
Investigative ophthalmology & visual science, 56(2), 1274-1282 (2015-01-31)
Previously, aliphatic β-nitroalcohols (BNAs) have been studied as a means to chemically induce tissue cross-linking (TXL) of cornea and sclera. There are a number of related and possibly more potent agents, known as formaldehyde releasers (FARs), that are in commercial

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service