Skip to Content
Merck
All Photos(2)

Key Documents

416908

Sigma-Aldrich

B-Iodo-9-BBN solution

1.0 M in hexanes

Synonym(s):

9-Iodo-9-borabicyclo[3.3.1]nonane solution

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
€193.00

€193.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
€193.00

About This Item

Empirical Formula (Hill Notation):
C8H14BI
CAS Number:
Molecular Weight:
247.91
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

€193.00


Check Cart for Availability

Request a Bulk Order

form

liquid

concentration

1.0 M in hexanes

density

0.816 g/mL at 25 °C

SMILES string

IB1C2CCCC1CCC2

InChI

1S/C8H14BI/c10-9-7-3-1-4-8(9)6-2-5-7/h7-8H,1-6H2/t7-,8+

InChI key

YAYIUFDUYUYPJC-OCAPTIKFSA-N

General description

B-Iodo-9-BBN is a boron-based organic compound that is commonly used as a reagent in organic synthesis for the iodoboration of alkynes, enolboration, the demethylation of phenol methyl ethers, and synthesis of boron-containing compounds.

Application

B-Iodo-9-BBN solution (1.0 M in hexanes) can be used as a Lewis acid in the selective synthesis of δ-keto esters via a Michael-type addition reaction to α,β-unsaturated ketones.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-9.4 °F - closed cup

Flash Point(C)

-23 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service