Skip to Content
Merck
All Photos(1)

Key Documents

415936

Sigma-Aldrich

3,4-Dihydro-2(1H)-quinolinone

98%

Synonym(s):

Hydrocarbostyril

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9NO
CAS Number:
Molecular Weight:
147.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

165-167 °C (lit.)

SMILES string

O=C1CCc2ccccc2N1

InChI

1S/C9H9NO/c11-9-6-5-7-3-1-2-4-8(7)10-9/h1-4H,5-6H2,(H,10,11)

InChI key

TZOYXRMEFDYWDQ-UHFFFAOYSA-N

General description

Series of 3,4-dihydro-2(1H)-quinolinone derivatives having sigma-1 receptor (σ1R) antagonist activity have been synthesized. 3,4-dihydro-2((1)H)-quinolinones have been synthesized using N-(1′-alkoxy)cyclopropyl-2-haloanilines as starting reagent. Cyclopropane ring expansion in the presence of palladium catalyst is the major step involved in the synthesis. Library of 3,4-dihydro-2(1H)-quinolinones have been synthesized through the rearrangement of β-lactam intermediates on the solid-phase

Application

3,4-Dihydro-2(1H)-quinolinone may be employed as medium supplement in the culture medium of Pseudomonas ayucida during enrichment culture experiments.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takayuki Tsuritani et al.
Organic letters, 11(5), 1043-1045 (2009-02-06)
N-(1'-Alkoxy)cyclopropyl-2-haloanilines are transformed to 3,4-dihydro-2((1)H)-quinolinones via palladium-catalyzed cyclopropane ring expansion. The reaction tolerates a variety of functional groups such as ester, nitrile, ether, and ketone groups.
Fabio Del Bello et al.
Journal of medicinal chemistry, 63(11), 5763-5782 (2020-05-07)
A series of novel 1,4-dioxane analogues of the muscarinic acetylcholine receptor (mAChR) antagonist 2 was synthesized and studied for their affinity at M1-M5 mAChRs. The 6-cyclohexyl-6-phenyl derivative 3b, with a cis configuration between the CH2N+(CH3)3 chain in the 2-position and
Synthesis of 4-amino-3, 4-dihydro-2 (1H)-quinolinones via β-lactam intermediates on the solid-phase.
Pei Y, et al.
Tetrahedron Letters, 38(19), 3349-3352 (1997)
So Youn Park et al.
Behavioural brain research, 365, 133-140 (2019-03-10)
Cerebrovascular dysfunction is associated with cognitive impairment in vascular dementia patients. This study aimed to explore augmented improvement of cognition and memory by aripiprazole add-on for cilostazol treatment in vascular dementia model. Male C57BL/6 mice were subjected to BCAS, and
Selective removal of nitrogen from quinoline and petroleum by Pseudomonas ayucida IGTN9m.
Kilbane JJ, et al.
Applied and Environmental Microbiology, 66(2), 688-693 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service