All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C4H3N3O4
CAS Number:
Molecular Weight:
157.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
mp
223-225 °C (dec.) (lit.)
SMILES string
OC(=O)c1n[nH]cc1[N+]([O-])=O
InChI
1S/C4H3N3O4/c8-4(9)3-2(7(10)11)1-5-6-3/h1H,(H,5,6)(H,8,9)
InChI key
ZMAXXOYJWZZQBK-UHFFFAOYSA-N
General description
4-Nitro-3-pyrazolecarboxylic acid is a pyrazole derivative. Catalytic activity of bimetallic colloids, composed of silver nanoparticles partially coated with Pd clusters, adsorbed on 4-nitro-3-pyrazolecarboxylic acid, has been investigated by Surface-enhanced Raman scattering (SERS) spectroscopy. Effects of this pyrrazole on blood flow and its palladium hydrogenation has been studied.
Application
4-Nitro-3-pyrazolecarboxylic acid may be used as a starting reagent for the synthesis of library of pyrazole derivatives, potential A3 adenosine receptor (A3AR) antagonists.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
SERS investigation on 4-nitro-3-pyrazolecarboxylic acid adsorbed on palladium-doped silver nanoparticles.
Pergolese B, et al.
Vibrational Spectroscopy, 48(1), 107-112 (2008)
Hydrogenation on Granular Palladium-containing Catalysts: II. Hydrogenation of Nitroheterocyclic Compounds
Russ. J. Org. Chem., 38, 269-271 (2002)
Jing Wei et al.
Neurochemistry international, 55(7), 637-642 (2009-06-23)
Adenosine is known to act as a neuromodulator by suppressing synaptic transmission in the central and peripheral nervous system. A(3) adenosine receptor (A(3)AR) antagonists were recently considered as potential drugs for the treatment of cardiac ischemia and inflammation diseases. To
Bo Xuan et al.
Acta pharmacologica Sinica, 23(8), 705-712 (2002-07-31)
Effects of C-nitropyrazoles and C-nitroazoles on ocular blood flow and retinal function recovery after ischemia have been studied. The compounds were tested on ocular blood flow of ocular hypertensive (40 mmHg) rabbit eyes with colored microsphere technique. They were also
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service