Skip to Content
Merck
All Photos(1)

Key Documents

413429

Sigma-Aldrich

Dioctyl phenylphosphonate

95%

Synonym(s):

DOPP

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
€188.00

€188.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
€188.00

About This Item

Linear Formula:
C6H5P(O)[O(CH2)7CH3]2
CAS Number:
Molecular Weight:
382.52
Beilstein:
2948496
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€188.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.478 (lit.)

bp

207 °C/4 mmHg (lit.)

density

0.967 g/mL at 25 °C (lit.)

functional group

phosphonate

SMILES string

CCCCCCCCOP(=O)(OCCCCCCCC)c1ccccc1

InChI

1S/C22H39O3P/c1-3-5-7-9-11-16-20-24-26(23,22-18-14-13-15-19-22)25-21-17-12-10-8-6-4-2/h13-15,18-19H,3-12,16-17,20-21H2,1-2H3

InChI key

HAKMAMKAFTZXOZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dioctyl phenylphosphonate is a plasticizer.[1]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S S Hassan et al.
Talanta, 54(6), 1153-1161 (2008-10-31)
Two novel uranyl PVC matrix membrane sensors responsive to uranyl ion are described. The first sensor incorporates tris(2-ethylhexyl)phosphate (TEHP) as both electroactive material and plasticizer and sodium tetraphenylborate (NaTPB) as an ion discriminator. The sensor displays a rapid and linear
Optimisation of poly (vinyl chloride) matrix membrane ion-selective electrodes for ammonium ions.
Davies OG, et al.
Analyst, 113(3), 497-500 (1988)
Iron ion sensor based on functionalized ZnO nanorods.
Khun K, et al.
Electroanalysis, 24(3), 521-528 (2012)
Masaaki Habara et al.
Biosensors & bioelectronics, 19(12), 1559-1563 (2004-05-15)
The higher sensitivity for sweeteners can be achieved by newly developed lipid/polymer membranes. The membrane is composed of lipids such as phosphoric acid di-n-hexadecyl ester and tetradodecylammoniumbromid, and a plasticizer, dioctyl phenylphosphonate. As a result of changing electric charge of
K Oohira et al.
Biophysical chemistry, 61(1), 29-35 (1996-08-30)
Electric characteristics of two kinds of membranes in response to NaCl and quinine were theoretically studied; one membrane composed of polyvinyl chloride (PVC) and dioctylphenylphosphonate (DOPP), and the other a lipid/PVC/DOPP membrane containing PVC, DOPP and a negatively charged lipid.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service