Skip to Content
Merck
All Photos(1)

Documents

412430

Sigma-Aldrich

PAMAM dendrimer, ethylenediamine core, generation 3.5 solution

10 wt. % in methanol

Synonym(s):

PAMAM G3.5

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[NH2(CH2)2NH2]:(G=3.5);dendri PAMAM(NHCH2CH2COONa)64
CAS Number:
Molecular Weight:
12927.69
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

description

sodium carboxylate surface groups

Quality Level

form

solution

mol wt

generation 3.5

feature

core type ethylenediamine core (2-carbon core)

concentration

10 wt. % in methanol

no. Surface Groups

64

refractive index

n20/D 1.346

bp

65 °C

density

0.81 g/mL at 25 °C

storage temp.

2-8°C

Physical form

Contains 64 surface carboxylate groups

Legal Information

Manufactured by Dendritech®, Inc.
Dendritech is a registered trademark of Dendritech, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Replace volatile organic solvents with water due to environmental concerns, overcoming challenges in solubility.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service