Skip to Content
Merck
All Photos(2)

Key Documents

412252

Sigma-Aldrich

(Ethoxycarbonylmethyl)triphenylphosphonium bromide

98%

Synonym(s):

(Carboxymethyl)triphenylphosphonium bromide ethyl ester, NSC 60450

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€63.80

€63.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€63.80

About This Item

Linear Formula:
CH3CH2OCOCH2P(C6H5)3Br
CAS Number:
Molecular Weight:
429.29
Beilstein:
3581273
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

€63.80


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

reaction suitability

reaction type: C-C Bond Formation

mp

145-150 °C (dec.) (lit.)

functional group

ester
phosphine

SMILES string

[Br-].CCOC(=O)C[P+](c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C22H22O2P.BrH/c1-2-24-22(23)18-25(19-12-6-3-7-13-19,20-14-8-4-9-15-20)21-16-10-5-11-17-21;/h3-17H,2,18H2,1H3;1H/q+1;/p-1

InChI key

VJVZPTPOYCJFNI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Wittig olefination reactions for synthesis of tetramic acid derivatives
  • Synthesis of precursors for Bergman cyclization
  • Preparation of PPAR agonists derived from thalidomide-related LXR antagonists
  • Synthesis of benzoxaboroles with antimalarial activity
  • Michael acceptor based cysteine protease inhibitors
  • Mechanistic studies of the IspH reductase protein

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service