Skip to Content
Merck
All Photos(1)

Key Documents

411701

Sigma-Aldrich

1,4,5,6,7,7-Hexachloro-5-norbornene-2,3-dicarboxylic acid

99%

Synonym(s):

1,2,3,4,7,7-Hexachlorobicyclo[2.2.1]hept-2-ene-5,6-dicarboxylic acid, Hexachloroendomethylenetetrahydrophthalic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H4Cl6O4
CAS Number:
Molecular Weight:
388.84
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

solid

mp

239-242 °C (lit.)

SMILES string

OC(=O)C1C(C(O)=O)[C@]2(Cl)C(Cl)=C(Cl)[C@@]1(Cl)C2(Cl)Cl

InChI

1S/C9H4Cl6O4/c10-3-4(11)8(13)2(6(18)19)1(5(16)17)7(3,12)9(8,14)15/h1-2H,(H,16,17)(H,18,19)/t1?,2?,7-,8+

InChI key

DJKGDNKYTKCJKD-SBUZQEQSSA-N

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G M Decad et al.
Journal of toxicology and environmental health, 9(5-6), 911-920 (1982-05-01)
The absorption, distribution, and excretion of a highly chlorinated dicarboxylic acid, chlorendic acid, was studied in the male Fischer 344 rat. [14C]Chlorendic acid was absorbed after an oral dose of 7.7 mumol per kilogram of body weight. The distribution in
S C Hasmall et al.
Toxicology and applied pharmacology, 144(2), 287-293 (1997-06-01)
Most nongenotoxic hepatocarcinogens such as diethylhexyl phthalate (DEHP) and chlorendic acid (CEA) induce hepatocyte replicative DNA synthesis. However, not all mitogens are carcinogenic; in National Toxicology Program (NTP) studies 1,4-dichlorobenzene (DCB) was not hepatocarcinogenic in the rat despite the induction
Y P Dragan et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 16(3), 525-547 (1991-04-01)
Several pharmaceutical agents, manufacturing chemicals, and environmental contaminants were found to act primarily as promoting agents in an initiation-promotion paradigm. The phenotypic distribution of four enzyme markers--placental glutathione-S-transferase (PGST), gamma-glutamyl transpeptidase (GGT), canalicular ATPase (ATPase), and glucose-6-phosphatase (G6Pase)--was analyzed in
Chlorendic acid.
IARC monographs on the evaluation of carcinogenic risks to humans, 48, 45-53 (1990-01-01)
Chlorendic acid.
Report on carcinogens : carcinogen profiles, 12, 94-95 (2011-08-19)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service