Skip to Content
Merck
All Photos(2)

Key Documents

411485

Sigma-Aldrich

Diisopropyl malonate

ReagentPlus®, 99%

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€39.40

€39.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€39.40

About This Item

Linear Formula:
(CH3)2CHO2CCH2CO2CH(CH3)2
CAS Number:
Molecular Weight:
188.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€39.40


Check Cart for Availability

Request a Bulk Order

Quality Level

product line

ReagentPlus®

Assay

99%

form

liquid

refractive index

n20/D 1.412 (lit.)

bp

93-95 °C/12 mmHg (lit.)

density

0.991 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CC(C)OC(=O)CC(=O)OC(C)C

InChI

1S/C9H16O4/c1-6(2)12-8(10)5-9(11)13-7(3)4/h6-7H,5H2,1-4H3

InChI key

QRVSDVDFJFKYKA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Diisopropyl malonates may be used in the synthesis of :
  • 3-substituted coumarins[1]
  • 2-carboisopropoxy-3-hydroxyquinoxaline-di-N-oxide[2]
  • 2-carboisopropoxy-3-hydroxy-6-methoxylquinoxaline-di-N-oxide[2]

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Catalytic Enantioselective Michael Addition of a Simple Malonate to Prochiral a, ?-Unsaturated Ketoses and Aldehydes.
Yamaguchi M, et al.
Angewandte Chemie (International Edition in English), 32(8), 1176-1178 (1993)
Yingjun Xu et al.
Molecules (Basel, Switzerland), 16(8), 6894-6901 (2011-08-16)
A series of quinoxaline 1,4-di-N-oxide analogues were prepared from benzofurazan N-oxide derivatives and β-diketone ester compounds by the improved Beirut reaction. The structures of the target products were characterized by NMR, MS, IR and elemental analysis measurements, and that of
H Jinno et al.
Archives of toxicology, 71(9), 550-555 (1997-01-01)
The fungicide isoprothiolane (diisopropyl 1,3-dithiolane-2-ylidenemalonate) decomposes to the diisopropyl esters of malonic acid (DM), chloromalonic acid (DCM) and dichloromalonic acid (DDCM) upon aqueous chlorination. In this study, the cytotoxicity of these compounds was examined using rat hepatocytes cultured on Matrigel.
Use of isopropyl alcohol as a solvent in Ti (O-i-Pr) 4-catalyzed Knoevenagel reactions.
Yamashita K, et al.
Tetrahedron, 61(33), 7981-7985 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service