Skip to Content
Merck
All Photos(2)

Key Documents

410470

Sigma-Aldrich

5-Bromo-2-hydroxy-3-methoxybenzaldehyde

97%

Synonym(s):

5-Bromo-3-methoxysalicylaldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€242.00

€242.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€242.00

About This Item

Linear Formula:
BrC6H2(OH)(OCH3)CHO
CAS Number:
Molecular Weight:
231.04
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€242.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

mp

125-127 °C (lit.)

functional group

aldehyde
bromo

SMILES string

COc1cc(Br)cc(C=O)c1O

InChI

1S/C8H7BrO3/c1-12-7-3-6(9)2-5(4-10)8(7)11/h2-4,11H,1H3

InChI key

MMFKBTPDEVLIOR-UHFFFAOYSA-N

Application

5-Bromo-2-hydroxy-3-methoxybenzaldehyde may be employed for the following syntheses:
  • ailanthoidol, via Stille coupling[1]
  • 6-bromo-8-methoxy-3-(methoxycarbonyl)-2H-chromen-2-one[2]
  • benzimidazole-based ligand, 2-(1H-benzoimidazol-2-yl)-4-bromo-6-methoxy-phenol (HL)[3]
  • chromogenic reagent, 5-bromo-2-hydroxy-3-methoxybenzaldehyde-p-hydroxy benzoic hydrazone[4]
  • (E)-N′-(5-bromo-2-hydroxy-3-methoxybenzylidene)-2-hydroxybenzohydrazide monohydrate[5]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shun-Yu Lin et al.
The Journal of organic chemistry, 68(7), 2968-2971 (2003-03-29)
Ailanthoidol 1, which can be isolated from Chinese herbal medicine, is achieved in which the longest linear sequence is only six steps in 48% overall yield from commercially available 5-bromo-2-hydroxy-3-methoxybenzaldehyde. The key transformations in the synthesis are the Stille coupling
Transformation of a luminescent benzimidazole-based Yb3 cluster into a one-dimensional coordination polymer.
Yang X, et al.
Crystal Growth & Design, 10(2), 970-976 (2009)
Direct Spectrophotometric Determination of Ni (II) Using 5-Bromo-2-hydroxyl-3-methoxybenzaldehyde-4-hydroxy benzoichydrazone.
Saritha B and Reddy TS.
J. Appl. Chem., 7(3), 22-26 (2014)
(E)-N'-(5-Bromo-2-hydroxy-3-methoxybenzylidene)-2-hydroxybenzohydrazide monohydrate.
Zhao S, et al.
Acta Crystallographica Section E, Structure Reports Online, 68(7), 2040-2040 (2012)
Pedro Verdía et al.
Molecules (Basel, Switzerland), 16(6), 4379-4388 (2011-05-31)
The ionic liquid 1,3-dimethylimidazolium methyl sulfate, [MMIm][MSO₄], together with a small amount of water (the amount taken up by the ionic liquid upon exposure to air), acts efficiently as both solvent and catalyst of the Knoevenagel condensation reactions of malononitrile

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service