Skip to Content
Merck
All Photos(1)

Key Documents

405175

Sigma-Aldrich

IR-1048

Dye content 97 %

Synonym(s):

1-Butyl-2-[2-[3-[(1-butyl-6-chlorobenz[cd]indol-2(1H)-ylidene)ethylidene]-2-chloro-1-cyclohexen-1-yl]ethenyl]-6-chlorobenz[cd]indolium tetrafluoroborate

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€278.00

€278.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€278.00

About This Item

Empirical Formula (Hill Notation):
C40H38BCl3F4N2
CAS Number:
Molecular Weight:
739.91
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

€278.00


Check Cart for Availability

Request a Bulk Order

composition

Dye content, 97%

Quality Level

mp

230 °C (dec.) (lit.)

λmax

1048 nm in ethanol

storage temp.

−20°C

SMILES string

F[B-](F)(F)F.CCCCN1/C(=C\C=C2/CCCC(\C=C\C3=[N+](CCCC)c4ccc(Cl)c5cccc3c45)=C2Cl)c6cccc7c(Cl)ccc1c67

InChI

1S/C40H38Cl3N2.BF4/c1-3-5-24-44-34(30-14-8-12-28-32(41)18-22-36(44)38(28)30)20-16-26-10-7-11-27(40(26)43)17-21-35-31-15-9-13-29-33(42)19-23-37(39(29)31)45(35)25-6-4-2;2-1(3,4)5/h8-9,12-23H,3-7,10-11,24-25H2,1-2H3;/q+1;-1

InChI key

IKZNCEDZIQWVNS-UHFFFAOYSA-N

Application

  • pH-Responsive Theranostic Colloidosome Drug Carriers: This study presents pH-responsive theranostic drug carriers for targeted thrombolytic therapy using the near-infrared-II window. It highlights the real-time imaging capabilities of these carriers using IR-1048, making it significant for drug delivery research (Y Ye, Z Chen, S Zhang, et al., 2022).
  • Targeted Nanobody Complex Enhanced Photodynamic Therapy: The article explores targeted therapy for lung cancer using IR-1048, which serves as a NIR-II/photoacoustic signal reporter. This study is pertinent for drug discovery chemists focusing on cancer treatment (Q Zhang, L Wu, S Liu, et al., 2020).

Features and Benefits

Q-switching IR laser dye.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service