Skip to Content
Merck
All Photos(1)

Documents

404810

Sigma-Aldrich

N,N′-Bis(3-aminopropyl)-1,3-propanediamine

technical grade, 90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2N(CH2)3NH(CH2)3NH(CH2)3NH2
CAS Number:
Molecular Weight:
188.31
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

90%

form

liquid

refractive index

n20/D 1.491 (lit.)

bp

98-103 °C/1 mmHg (lit.)

density

0.92 g/mL at 25 °C (lit.)

SMILES string

NCCCNCCCNCCCN

InChI

1S/C9H24N4/c10-4-1-6-12-8-3-9-13-7-2-5-11/h12-13H,1-11H2

InChI key

ZAXCZCOUDLENMH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N′-Bis(3-aminopropyl)-1,3-propanediamine, a linear polyamine, has been identified in white shrimp Penaeus setiferus by gas chromatography-mass spectrometry.

Application

N,N′-Bis(3-aminopropyl)-1,3-propanediamine (bappn) may be used in the preparation of [Ni(bappn)(ttcH)]·5H2O complex (ttch= trithiocyanurate dianion).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

R H Hu et al.
The Biochemical journal, 328 ( Pt 1), 307-316 (1998-01-10)
Treatment of Chinese hamster ovary cells with alpha-difluoromethylornithine for 3 days, followed by exposure to cycloheximide, led to an unregulated, rapid and massive accumulation of polyamine analogues. This accumulation led to cell death by apoptosis within a few hours. Clear
Identification of the unusual polyamines 3,3'-diaminodipropylamine and N,N'-bis(3-aminoproply)-1,3-propanediamine in the white shrimp Penaeus setiferus.
L W Stillway et al.
Biochemical and biophysical research communications, 77(3), 1103-1107 (1977-08-08)
K Hamana et al.
Journal of biochemistry, 91(4), 1321-1328 (1982-04-01)
Seven phyla of eukaryotic algae were analyzed to determine their contents of diamines and polyamines. The algae examined included Rhodophyta, Pyrrophyta, Chrysophyta, Phaeophyta, Euglenophyta, Chlorophyta, and Charophyta. Both putrescine and spermidine were detected in all the algae studied, while appreciable
Anurupa Shrestha et al.
Bioorganic & medicinal chemistry letters, 19(9), 2478-2481 (2009-04-01)
We have previously shown that simple N-acyl or N-alkyl polyamines bind to and sequester Gram-negative bacterial lipopolysaccharide, affording protection against lethality in animal models of endotoxicosis. Several iterative design-and-test cycles of SAR studies, including high-throughput screens, had converged on compounds
Andrew J Geall et al.
Bioconjugate chemistry, 13(3), 481-490 (2002-05-16)
Polyamine amides have been prepared from lithocholic and cholic acids (5beta-colanes) by acylation of tri-Boc-protected tetraamines spermine and thermine. These designed ligands for DNA are polyammonium ions at physiological pH. In NMR spectra, they display 14N-1H 1J = 51 Hz

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service