Skip to Content
Merck
All Photos(1)

Key Documents

40296

Sigma-Aldrich

Methanol

Synonym(s):

Methyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OH
CAS Number:
Molecular Weight:
32.04
Beilstein:
1098229
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

vapor density

1.11 (vs air)

vapor pressure

410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)

form

liquid

CofA

specification on request

autoignition temp.

725 °F

expl. lim.

36 %

refractive index

n20/D 1.329 (lit.)

bp

64.7 °C (lit.)

mp

−98 °C (lit.)

density

0.791 g/mL at 25 °C (lit.)

SMILES string

CO

InChI

1S/CH4O/c1-2/h2H,1H3

InChI key

OKKJLVBELUTLKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Semiconductor grade methanol may be used to degas indium antimonide wafers. Poly(amidoamine) (PAMAM) was coupled with carboxylic acid terminated Si surfaces by soaking the Si surfaces in methanol. Methanol may be used to degrease silicon ingots and ribbons. It may be used in the purification of surfactant (sodium diethylhexylsulfosuccinate) and to prepare a colloidal suspension of silicon.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

LA-ICP-MS, IC and DPASV-DPCSV determination of metallic impurities in solar-grade silicon.
Buldini PL, et al.
Talanta, 47(1), 203-212 (1998)
Luminescent colloidal silicon suspensions from porous silicon.
Heinrich JL, et al.
Science, 255, 66-68 (1992)
Investigation of the mobility of amphiphilic polymer?AOT reverse microemulsion systems using electron spin resonance
Wines TH, et al.
Journal of Colloid and Interface Science, 285(1), 318-325 (2005)
Growth and characterization of indium antimonide and gallium antimonide crystals.
Udayashankar NK and Bhat HL
Bulletin of Materials Science, 24(5), 445-453 (2001)
Immobilization of dendrimers on Si?C linked carboxylic acid-terminated monolayers on silicon (111).
Bocking T, et al.
Thin Solid Films, 515(4), 1857-1863 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service