Skip to Content
Merck
All Photos(1)

Key Documents

401552

Sigma-Aldrich

Calcium pyrophosphate

≥99.9% trace metals basis

Synonym(s):

Dicalcium pyrophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ca2P2O7
CAS Number:
Molecular Weight:
254.10
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Assay

≥99.9% trace metals basis

form

powder

mp

1353 °C (lit.)

solubility

H2O: insoluble(lit.)

density

3.09 g/mL at 25 °C (lit.)

SMILES string

[Ca++].[Ca++].[O-]P([O-])(=O)OP([O-])([O-])=O

InChI

1S/2Ca.H4O7P2/c;;1-8(2,3)7-9(4,5)6/h;;(H2,1,2,3)(H2,4,5,6)/q2*+2;/p-4

InChI key

JUNWLZAGQLJVLR-UHFFFAOYSA-J

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongyi He et al.
BMC musculoskeletal disorders, 21(1), 97-97 (2020-02-14)
Patients with chondrocalcinosis may suffer from a series of symptoms resembling acute gouty arthritis or septic arthritis, but the aetiology and pathogenesis of chondrocalcinosis have not been fully elucidated yet. This study was aimed to assess serum zinc and copper
Anna Moltó et al.
Joint, bone, spine : revue du rhumatisme, 79(6), 621-623 (2012-06-05)
Acute synovitis induced by deposition of calcium pyrophosphate (CPP) and monosodium urate crystals involves interleukin-1β production and activation. The efficacy of blocking interleukin-1β activity (with an interleukin-1 receptor antagonist [anakinra] or interleukin-1β antibody) is well documented for gout attacks but
Sébastien Ottaviani et al.
Joint, bone, spine : revue du rhumatisme, 80(2), 178-182 (2012-10-02)
Calcium pyrophosphate (CPP) crystal-induced arthritis occurs particularly in elderly people. This population has frequently associated comorbidities and treatments, which could limit the use of conventional therapies (colchicine, non-steroidal anti-inflammatory drugs and corticosteroids). The aim of the study was to evaluate
Torsten Diekhoff et al.
Investigative radiology, 50(4), 255-260 (2014-10-22)
The aim of this study was to perform phantom measurements to prove the feasibility of single-source dual-energy computed tomography (DECT) of the extremities using a volume scan mode. In addition, we, for the first time, wanted to determine which concentrations
Yi-Jie Kuo et al.
PloS one, 7(7), e40272-e40272 (2012-07-14)
Treatment for osteoporosis commonly includes the use of bisphosphonates. Serious side effects of these drugs are caused by the inhibition of bone resorption as a result of osteoclast apoptosis. Treatment using calcitonin along with bisphosphonates overcomes these side-effects in some

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service