Skip to Content
Merck

401129

Cadmium tungstate

powder, −325 mesh

Synonym(s):

CWO, Cadmium tungsten oxide

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Linear Formula:
CdWO4
CAS Number:
Molecular Weight:
360.25
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Cadmium tungstate, −325 mesh

SMILES string

[Cd++].[O-][W]([O-])(=O)=O

InChI

1S/Cd.4O.W/q+2;;;2*-1;

InChI key

OUMLAYUZCYYIOD-UHFFFAOYSA-N

form

powder

reaction suitability

core: cadmium, reagent type: catalyst

particle size

−325 mesh

Application

Used for:
  • Preparation of CdWO4 polymorphs to study electronic structures and photocatalytic activities
  • Scintillating bolometers for double beta decay search
  • Studies of laser photochemical ablation

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

7790-85-4

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Iwanaga et al.
International journal of cosmetic science, 42(3), 229-236 (2020-01-30)
Particulate matter (PM), such as air pollutants and pollens, are known to cause skin ageing through skin inflammation. It is important to develop formulations which protect the skin from PM. We previously developed a conventional water-in-oil emulsion with a synthetic
P F Kirvan et al.
Medical physics, 37(1), 249-257 (2010-02-24)
Megavoltage computed tomography (MVCT) and megavoltage cone beam computed tomography (MVCBCT) can be used for visualizing anatomical structures prior to radiation therapy treatments to assist in patient setup and target localization. These systems are less susceptible to metal artifacts and
Prejaas Tewarie et al.
Annals of clinical and translational neurology, 4(9), 614-621 (2017-09-15)
We conducted a multi-modal optical coherence tomography (OCT) and magnetoencephalography (MEG) study to test whether there is a relationship between retinal layer integrity and electrophysiological activity and connectivity (FC) in the visual network influenced by optic neuritis (ON) in patients
Junpeng Zhang et al.
Environmental pollution (Barking, Essex : 1987), 246, 131-140 (2018-12-12)
Bacterial antibiotic resistance (BAR) is profoundly important to human health, but the environmental reservoirs of resistance determinants are poorly understood. BAR of biofilm and tap water were analyzed by using a water distribution simulator where different doses of chlorine and
S L Fritz et al.
Medical physics, 14(2), 244-248 (1987-03-01)
A high-resolution x-ray detector array for x-ray imaging is described. It consists of a discrete array of CdWO4 scintillators and a Reticon 1024S photodiode array with a fiber-optic faceplate. The scintillator elements measure 0.099 X 3 X 4 mm and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service