Skip to Content
Merck
All Photos(1)

Key Documents

399760

Sigma-Aldrich

4,5-Difluoroanthranilic acid

97%

Synonym(s):

2-Amino-4,5-difluorobenzoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€58.30

€58.30


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
€58.30

About This Item

Linear Formula:
F2C6H2(NH2)CO2H
CAS Number:
Molecular Weight:
173.12
Beilstein:
2834162
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

€58.30


Estimated to ship on29 May 2025


Request a Bulk Order

Assay

97%

reaction suitability

reaction type: solution phase peptide synthesis

mp

181-183 °C (lit.)

application(s)

peptide synthesis

SMILES string

Nc1cc(F)c(F)cc1C(O)=O

InChI

1S/C7H5F2NO2/c8-4-1-3(7(11)12)6(10)2-5(4)9/h1-2H,10H2,(H,11,12)

InChI key

DGOZIZVTANAGCA-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service