Skip to Content
Merck
All Photos(1)

Documents

399701

Sigma-Aldrich

(R)-(+)-1-Octyn-3-ol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH(OH)C≡CH
CAS Number:
Molecular Weight:
126.20
MDL number:
UNSPSC Code:
12352003
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

optical activity

[α]20/D +6.5°, c = 2 in methylene chloride

optical purity

ee: 98% (GLC)

refractive index

n20/D 1.442 (lit.)

bp

100 °C/20 mmHg (lit.)

density

0.864 g/mL at 25 °C (lit.)

SMILES string

CCCCC[C@@H](O)C#C

InChI

1S/C8H14O/c1-3-5-6-7-8(9)4-2/h2,8-9H,3,5-7H2,1H3/t8-/m0/s1

InChI key

VUGRNZHKYVHZSN-QMMMGPOBSA-N

Application

Key intermediate for the synthesis of prostaglandins.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Organic Chemistry, 53, 5590-5590 (1988)
Journal of the American Chemical Society, 110, 4718-4718 (1988)
Tetrahedron Letters, 30, 3645-3645 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service