Skip to Content
Merck
All Photos(1)

Key Documents

395315

Sigma-Aldrich

Ethyl diazoacetoacetate

Synonym(s):

Ethyl Α-diazoacetoacetate, Ethyl 2-diazo-3-oxobutanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COC(N2)CO2C2H5
CAS Number:
Molecular Weight:
156.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.36 psi ( 20 °C)

form

liquid

availability

available only in USA

refractive index

n20/D 1.474 (lit.)

density

1.131 g/mL at 25 °C (lit.)

functional group

ester
ketone

SMILES string

CCOC(=O)C(=[N+]=[N-])C(C)=O

InChI

1S/C6H8N2O3/c1-3-11-6(10)5(8-7)4(2)9/h3H2,1-2H3

InChI key

JWTPSIXYXYNAOU-UHFFFAOYSA-N

General description

Ethyl diazoacetoacetate is a diazo compound. It participates in Rh2(OAc)4-catalyzed reactions with arylalkylamines and diarylamines.[1] Reaction of ethyl diazoacetoacetate with N-methyl pyrrole and pyrrole derivatives has been studied.[2]

Application

Ethyl diazoacetoacetate can be used as a reactant to synthesize:
  • 1,4-oxathiocines and thiopyran derivatives via Rh-catalyzed reaction with 2-amino-4,5-dihydro-3-thiophenecarbonitriles.[3]
  • β-keto esters via C−H insertion reaction with aromatic aldehydes using NbCl5 as a catalyst.[4]
  • Diazoacetoacetate derivatives by reacting with aldehydes via aldol condensation and subsequent and in situ oxidation reaction.[5]
  • Isoquinolone and pyridone derivatives by Rh-catalyzed C−H activation/annulation reaction with various N-methoxybenzamides.[6]

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Self-react. C - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Niobium (V) chloride-catalyzed C-H insertion reactions of a-diazoesters: synthesis of ?-keto esters.
Yadav JS, et al.
Tetrahedron, 61(4), 875-878 (2005)
Reaction of dimethyl diazomalonate and ethyl 2-diazoacetoacetate with N-methylpyrrole.
Maryanoff BE.
The Journal of Organic Chemistry, 47(15), 3000-3002 (1982)
Niobium (V) chloride-catalyzed C-H insertion reactions of ?-diazoesters: synthesis of β-keto esters
Yadav JS, et al.
Tetrahedron, 61(4), 875-878 (2005)
Synthesis of 1, 4-Oxathiocines and Thiopyrans by the Reaction of 2-Amino-4, 5-dihydro-3-thiophenecarbonitriles with Ethyl Diazoacetoacetate
Yamagata K, et al.
Liebigs Annalen der Chemie , 1996(5), 725-729 (1996)
M Yang et al.
The Journal of organic chemistry, 66(20), 6729-6733 (2001-10-02)
The Rh(2)(OAc)(4)-stabilized carbenoid derived from dimethyl diazomalonate has been found to insert into the N-H bond of sterically hindered secondary aliphatic amines to afford hindered tertiary aliphatic amines in quite satisfactory yields. For example dimethyl 2-(dicyclohexylamino)propanedioate was formed in 85%

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service