Skip to Content
Merck
All Photos(1)

Key Documents

394580

Sigma-Aldrich

Myristic anhydride

95%

Synonym(s):

Tetradecanoic anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)12CO]2O
CAS Number:
Molecular Weight:
438.73
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

53-55 °C (lit.)

functional group

anhydride
ester

SMILES string

CCCCCCCCCCCCCC(=O)OC(=O)CCCCCCCCCCCCC

InChI

1S/C28H54O3/c1-3-5-7-9-11-13-15-17-19-21-23-25-27(29)31-28(30)26-24-22-20-18-16-14-12-10-8-6-4-2/h3-26H2,1-2H3

InChI key

RCRYHUPTBJZEQS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Myristic anhydride is an anhydride of myristic acid. Its synthesis by reacting myristoyl chloride with acetic anhydride has been reported.[1] It participates in the synthesis of fatty acyl derivatives of macrocyclic peptides.[2]

Application

Myristic anhydride may be used in the following studies:
  • As a reactant to synthesize cholesteryl myristate by reacting with cholesterol.[3]
  • As a reactant in the synthesis of 1,2-dimyristoyl-sn-glycero-3-phosphocholine by acylating sn-glycero-3-phosphocholine.[4]
  • As a reagent in the synthesis of 3-(bis-{2-(tert-butyldimethylsilyloxy)ethyl}amino)propane-1,2-diol, an intermediate of cationic lipid.[5]
  • As a reactant to synthesize neryl myristate by reacting with nerol.[6]
  • As a reagent to synthesize myristoylated t-butylglutamic acid by myristoylation of γ-tert-butoxy-glutamic acid.[7]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of saturated, unsaturated, spin-labeled, and fluorescent cholesteryl esters: Acylation of cholesterol using fatty acid anhydride and 4-pyrrolidinopyridine.
Patel KM, et al.
Lipids, 14(9), 816-818 (1979)
Bhupender S Chhikara et al.
Tetrahedron letters, 53(39), 5335-5337 (2012-11-24)
Designing microbicidal gels of anti-HIV drugs for local application to prevent HIV infection is a subject of major interest. 3'-Fluoro-3'-deoxythymidine (FLT), a nucleoside reverse transcriptase inhibitor (NRTI), was conjugated with a N-myristoyl glutamate scaffold. The conjugate showed gelation at 1%
Michael H Nantz et al.
Molecular pharmaceutics, 7(3), 786-794 (2010-03-18)
We, and others, have observed that the structure of cationic lipids appears to have a significant effect on the transfection efficacy of optimized nucleic acid/cationic lipid complexes (lipoplexes) used for in vitro and in vivo gene delivery and expression. Although
Reactions of fatty acid chlorides. I. Preparation of fatty acid anhydrides.
Sonntag NOV, et al.
Journal of the American Oil Chemists' Society, 31(4), 151-157 (1954)
Neryl myristate from the acarid mite, Aleuroglyphus ovatus (acarina, acaridae).
Leal WS, et al.
Agricultural and Biological Chemistry, 52(5), 1299-1300 (1988)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service