Skip to Content
Merck
All Photos(2)

Key Documents

394378

Sigma-Aldrich

4-Bromo-2,6-di-tert-butylphenol

98%

Synonym(s):

2,6-Di-tert-butyl-4-bromophenol, 4-Bromo-2,6-bis(1,1-dimethylethyl)phenol, 4-Hydroxy-3,5-di(tert-butyl)bromobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H2[C(CH3)3]2OH
CAS Number:
Molecular Weight:
285.22
Beilstein:
2051052
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

78-83 °C (dec.) (lit.)

functional group

bromo

SMILES string

CC(C)(C)c1cc(Br)cc(c1O)C(C)(C)C

InChI

1S/C14H21BrO/c1-13(2,3)10-7-9(15)8-11(12(10)16)14(4,5)6/h7-8,16H,1-6H3

InChI key

SSQQUEKFNSJLKX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Bromo-2,6-di-tert-butylphenol is a 4-substituted-2,6-di-tert-butylphenol. It is synthesized by the bromination of 2,6-di-tert-butylphenol. The structure was confirmed by 1H NMR.[1] Its photolysis reaction in benzene solution has been investigated.[2] The photochemical reaction of 4-bromo-2,6-di-tert-butylphenol single crystals doped with 2,6-di-tert-butyl-p-quinone has been studied by EPR spectroscopy.[3]

Application

4-Bromo-2,6-di-tert-butylphenol may be used in the following studies:
  • As a terminating comonomer phenol in the phase transfer catalyzed (PTC) polymerization of 4-bromo-2,6-dimethylphenol.[1]
  • Synthesis of 1,1-[1,10-decanediylbis(oxy)]bis[(2,6-ditertbutyl-4-bromo)benzene], a monomer, which forms poly(p-phenylenevinylene) derivatives by reaction with 1,10-dibromodecane.[4]
  • As a reactant in the synthesis of 2,6-di-tert-butyl-phenolnorbornene (NArOH), a norbornene comonomer bearing an antioxidant hindered phenol.[5]
  • As a catalyst with methyl aluminium to form methylaluminum bis(4-bromo-2,6-di-tert-butylphenoxide) (MABR) which may be utilized for the transformation of various epoxides to carbonyl compounds.[6][7]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Blue light-emitting poly (p-phenylenevinylene) derivatives containing alternating conjugated segments and aliphatic spacers.
Mpallas JG, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 41(8), 1091-1098 (2003)
An efficient, catalytic procedure for epoxide rearrangement.
Maruoka K, et al.
Tetrahedron Letters, 30(41), 5607-5610 (1989)
Effect of donor and acceptor on the mechanism of a photochemical reaction in doped single crystals for radical pairs in 4-bromo-2, 6-di-tert-butylphenol single crystal doped with 2, 6-di-tert-butyl-p-quinone.
Tipikin DS, et al.
Kinetics and Catalysis, 42(2), 246-250 (2001)
Phase transfer catalyzed polymerization of 4-bromo-2, 6-dimethylphenol in the presence of a terminating comonomer phenol: 2, 4, 6-tri-tert-butylphenol or 4-bromo-2, 6-di-tert-butylphenol.
Wang JH and Percec V.
Polym. Bull., 25(1), 33-40 (1991)
Photolysis of 4-bromo-2, 6-di-tert-butylphenol in benzene solution.
Lappin GR and Zannucci JS.
Tetrahedron Letters, 10(58), 5085-5087 (1969)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service