Skip to Content
Merck
All Photos(1)

Documents

392146

Sigma-Aldrich

4-Imidazoledithiocarboxylic acid

70%, technical grade

Synonym(s):

5-Imidazoledithiocarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4N2S2
CAS Number:
Molecular Weight:
144.22
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

grade

technical grade

Assay

70%

mp

145 °C (dec.) (lit.)

SMILES string

SC(=S)c1c[nH]cn1

InChI

1S/C4H4N2S2/c7-4(8)3-1-5-2-6-3/h1-2H,(H,5,6)(H,7,8)

InChI key

RAUHREXYGRKIOJ-UHFFFAOYSA-N

General description

4-Imidazoledithiocarboxylic acid is an imidazole derivative.

Application

4-Imidazoledithiocarboxylic acid (4(5)-imidazoledithiocarboxylic acid, imidazole-4-dithiocarboxylic acid) is suitable for use in the preparation of imidazole functionalized silica with enhanced metal uptake and 3H-imidazole-4-carbodithioic acid 4-vinyl-benzyl ester, a RAFT (reversible addition-fragmentation chain transfer) agent used in the synthesis of highly-branched polymers.
It may be used in the synthesis of the following:
  • Nucleophilic fatty acid ligands containing imidazole derivatives.
  • Diorganotin (IV) complexes by reacting with diorganotin (IV) dichlorides.
  • 2-(Imidazol-4-yl-4′)benzimidazole by reacting with o-phenylenediamine.

Packaging

5 g in glass bottle

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reactive silver and cobalt nanoparticles modified with fatty acid ligands functionalized by imidazole derivatives.
Bromberg L, et al.
Chemistry of Materials, 22(18), 5383-5391 (2010)
Azolylimidazoles: Synthetic strategies and medicinal applications.
Turkish Journal of Chemistry 38.1 (2014): 1-27.
Abdelwahab BF and Khidre RE.
Turkish Journal of Chemistry, 38(1), 1-27 (2014)
Highly-branched SMART polymers using styryl dithioate esters.
Carter SR, et al.
Polymer Preprints (American Chemical Society, Division of Polymer Chemistry), 46(2), 253-253 (2005)
Functionalized SiO2 with N-, S- containing ligands for Pb (II) and Cd (II) adsorption.
Grigoropoulou G, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 320(1), 25-35 (2008)
Investigation on the coordination modes: Syntheses, characterization and crystal structures of diorganotin (IV) dichloride with 4 (5)-imidazoledithiocarboxylic acid.
Ma C, et al.
Inorgorganica Chimica Acta, 358(11), 5383-5391 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service