Skip to Content
Merck
All Photos(1)

Key Documents

391875

Sigma-Aldrich

(Dimethylaminomethylene)dimethylammonium chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCH=N(CH3)2Cl
CAS Number:
Molecular Weight:
136.62
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

130-139 °C (lit.)

storage temp.

2-8°C

SMILES string

[Cl-].[H]\C(N(C)C)=[N+](\C)C

InChI

1S/C5H13N2.ClH/c1-6(2)5-7(3)4;/h5H,1-4H3;1H/q+1;/p-1

InChI key

BPBLGCSAYMJJJW-UHFFFAOYSA-M

General description

(Dimethylaminomethylene)dimethylammonium chloride is an amidine derivative.

Application

(Dimethylaminomethylene)dimethylammonium chloride (N,N,N′,N′-tetramethylformamidinium chloride) may be used in the synthesis of the following:
  • aminomethylene hydantoins[1]
  • thiohydantoins[1]
  • 2,2′-o-phenyl­enebis(1,3-di­meth­ylguanidine)[2]
  • N,N-dimethylformamide di-tert-butyl acetal[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

2, 2'-o-Phenylenebis (1,3-dimethylguanidine).
Kawahata M, et al.
Acta Crystallographica Section E, Structure Reports Online, 62(8), 3301-3302 (2006)
Synthesis of 5-chloromethylene hydantoins and thiohydantoins.
Cernak T A and Gleason J L.
Heterocycles, 71(1), 117-134 (2007)
Robert HD
Carboxylic Ortho Acid Derivatives: Preparation and Synthetic Applications (Academic press), 14, 423-423 null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service