Skip to Content
Merck
All Photos(1)

Key Documents

391468

Sigma-Aldrich

4-Hydroxypiperidine hydrochloride

98%

Synonym(s):

4-Piperidinol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11NO · HCl
CAS Number:
Molecular Weight:
137.61
Beilstein:
3908995
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

155-157 °C (lit.)

solubility

methanol: soluble 25 mg/mL, clear, colorless

functional group

hydroxyl

SMILES string

Cl[H].OC1CCNCC1

InChI

1S/C5H11NO.ClH/c7-5-1-3-6-4-2-5;/h5-7H,1-4H2;1H

InChI key

VKCORPXOKYDINR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Hydroxypiperidine hydrochloride is a piperidine derivative. The synthesis of 4-hydroxypiperidine has been reported. 4-Hydroxypiperidine participates in the synthesis of 4-(4′-hydroxypiperidino)-4-phenylcyclohexanol and 1-(4-hydroxypiperidino)cyclohexanecarbonitrile. 4-Hydroxypiperidine is the starting material for the synthesis of Sibrafiban and Lamifiban, two non-peptidefibrinogen receptor.

Application

4-Hydroxypiperidine hydrochloride is suitable for use in the Ugi-multi component reaction (MCR) to study the use of miniaturised-synthesis and Total Analysis System (mSYNTAS) in solution phase synthesis and on-line analysis of Ugi-MCRs. It may be used in the synthesis of acetic acid-piperidine-4-yl ester.
Reactant for synthesis of:
14C-labeled E4010
PET tracer for imaging brain acetylcholinesterase

Reactant for allosteric modulators of metabotropic glutamate receptor subtype 5

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Synthesis of Δ3-Tetrahydropyridine.
Renshaw RR and Conn RC.
Journal of the American Chemical Society, 60(4), 745-747 (1938)
Synthesis of 4-(4'-hydroxypiperidino)-4-phenylcyclohexanol, a dihydroxy phencyclidine metabolite.
Brine GA, et al.
Organic preparations and procedures international, 15(6), 371-377 (1983)
Concise Synthesis of Sibrafiban and Lamifiban, Two Non-Peptide Fibrinogen Receptor (GPIIb/IIIa) Antagonists.
Chang MY and Chen ST.
J. Chin. Chem. Soc., 48(2), 133-135 (2001)
M C Mitchell et al.
The Analyst, 126(1), 24-27 (2001-02-24)
A miniaturised-SYNthesis and Total Analysis System (mu SYNTAS) was used for the solution-phase synthesis and on-line analysis (TOF-MS) of Ugi multicomponent reaction (MCR) products. This approach provides an unusually high degree of control of the MCR and delivers detailed, novel
C Deleuze-Masquefa et al.
Journal of medicinal chemistry, 40(25), 4019-4025 (1997-12-24)
1-[1-(2-Benzo[b]thiopheneyl)cyclohexyl]piperidine hydrochloride (BTCP, 1) and cocaine bind to the neuronal dopamine transporter to inhibit dopamine (DA) reuptake. However, on chronic administration, cocaine produces sensitization, but 1 produces tolerance. Because metabolites of 1 might be responsible for some of its pharmacological

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service