Skip to Content
Merck
All Photos(1)

Documents

388297

Sigma-Aldrich

Ferrocenium hexafluorophosphate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10F6FeP
CAS Number:
Molecular Weight:
331.00
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

reaction suitability

core: iron
reagent type: catalyst

SMILES string

[Fe+].[CH]1[CH][CH][CH][CH]1.[CH]2[CH][CH][CH][CH]2.F[P-](F)(F)(F)(F)F

InChI

1S/2C5H5.F6P.Fe/c2*1-2-4-5-3-1;1-7(2,3,4,5)6;/h2*1-5H;;/q;;-1;+1

InChI key

JTRCXNUQIANBAN-UHFFFAOYSA-N

Application

The ferrocenium cation has been used to prepare catalysts for the asymmetric epoxidation of olefins, carbonylations of nitroaromatics, and the isomerization of a strained diene.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giorgia Zampardi et al.
Physical chemistry chemical physics : PCCP, 19(41), 28381-28387 (2017-10-17)
High voltage operating active materials are among the most promising components for positive electrodes of future high energy lithium-ion batteries. However, the operating potential range of such materials often exceeds anodically the thermodynamic stability window of the electrolyte. A surface
Johanna Karppi et al.
Frontiers in chemistry, 8, 11-11 (2020-02-13)
Pyranose dehydrogenases (PDHs; EC 1.1.99.29; AA3_2) demonstrate ability to oxidize diverse carbohydrates. Previous studies of these enzymes have also uncovered substrate-dependent regioselectivity, along with potential to introduce more than one carbonyl into carbohydrate substrates. Enzymatic oxidation of carbohydrates facilitates their
Go Tajima et al.
Molecular genetics and metabolism, 119(4), 322-328 (2016-11-20)
Medium-chain acyl-CoA dehydrogenase (MCAD) deficiency is a representative disorder of fatty acid oxidation and is one of the most prevalent inborn errors of metabolism among Caucasian populations. In Japan, however, it was as late as 2000 when the first patient
Aifang Wang et al.
Journal of the American Chemical Society, 131(19), 6652-6653 (2009-05-01)
Redox-active ferrocenyl-terminated dendrimers have been used to build robust and active interfaces in which the inherent properties of the dendrimers are preserved. The procedure is based on the preparation of a polyarylcarboxylate layer through the controlled reduction of aryldiazonium salts
Krishnan Venkatasubbaiah et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 14(2), 444-458 (2007-12-12)
A series of mixed-valent (MV) complexes [(FeCp)2(mu-C10H6(BPh)2)]+X ([1+]X; X=I 5, PF6, SbF6, B(C6F5)4) were prepared by oxidation of diboradiferrocene [(FeCp)2(mu-C10H6(BPh)2)] (1) with I 2, AgPF6, and AgSbF6, respectively, and through anion exchange of the I 5(-) salt with [Li(Et2O)x][B(C6F5)4] in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service