Skip to Content
Merck
All Photos(2)

Key Documents

388173

Sigma-Aldrich

Titanium(IV) (triethanolaminato)isopropoxide solution

80 wt. % in isopropanol

Synonym(s):

TYZOR® TE organic titanate

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€40.60

€40.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€40.60

About This Item

Empirical Formula (Hill Notation):
C9H19NO4Ti
CAS Number:
Molecular Weight:
253.12
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

€40.60


Check Cart for Availability

Request a Bulk Order

form

viscous liquid

Quality Level

reaction suitability

core: titanium
reagent type: catalyst

concentration

80 wt. % in isopropanol

density

1.087 g/mL at 25 °C

SMILES string

CC(C)[Ti]12OCCN(CCO1)CCO2

InChI

1S/C6H12NO3.C3H7.Ti/c8-4-1-7(2-5-9)3-6-10;1-3-2;/h1-6H2;3H,1-2H3;/q-3;;+3

InChI key

RSUVVMHMKLWZQT-UHFFFAOYSA-N

Legal Information

TYZOR is a registered trademark of E. I. du Pont de Nemours and Company

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

60.8 °F

Flash Point(C)

16 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service