Skip to Content
Merck
All Photos(1)

Key Documents

387622

Sigma-Aldrich

Vinyl chloride

≥99.5%

Synonym(s):

Chloroethylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCl
CAS Number:
Molecular Weight:
62.50
Beilstein:
1731576
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:

Assay

≥99.5%

contains

either phenol or hydroquinone as inhibitor

refractive index

n20/D 1.3700 (lit.)

bp

-13.4 °C (lit.)

mp

-153.8 °C (lit.)

density

0.911 g/mL at 25 °C (lit.)

SMILES string

ClC=C

InChI

1S/C2H3Cl/c1-2-3/h2H,1H2

InChI key

BZHJMEDXRYGGRV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Recommended products

Stainless steel control valves Z146951, Z187224 or either of stainless steel regulators Z148504 and Z148512 and a "T"-purge valve Z151661 is recommended.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1A - Flam. Gas 1A - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

WGK 2

Flash Point(F)

-79.6 °F - closed cup

Flash Point(C)

-62 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ashish Thakur et al.
Chemical communications (Cambridge, England), 48(2), 203-205 (2011-11-16)
A protocol for the Suzuki-Miyaura coupling of heteroaryl boronic acids and vinyl chlorides that minimizes protodeboronation is described. A combination of catalytic amounts of Pd(OAc)(2) and SPhos in conjunction with CsF in isopropanol effectively affords a variety of coupled products.
Vinyl chloride monomer and hepatocarcinoma.
N Bourgeois
Acta gastro-enterologica Belgica, 64(1), 25-34 (2001-04-27)
R Lewis
Occupational medicine (Philadelphia, Pa.), 14(4), 719-742 (1999-09-25)
Polyvinyl chloride (PVC) is an important plastic resin for construction, pipe and tubing, siding, and other uses. Exposures to vinyl chloride monomer during the early years of production resulted in an important sentinel health event: the recognition of an excess
J Kielhorn et al.
Environmental health perspectives, 108(7), 579-588 (2000-07-25)
Vinyl chloride (VC) is both a known carcinogen and a regulated chemical, and its production capacity has almost doubled over the last 20 years, currently 27 million tons/year worldwide. According to recent reports it is still a cause for concern.
H J Clewell et al.
Chemosphere, 31(1), 2561-2578 (1995-07-01)
Risk assessments for vinyl chloride (VC) and trichloroethylene (TCE) are presented as examples of approaches for incorporating chemical-specific pharmacokinetic and mechanistic information into a more scientifically plausible cancer risk assessment. For VC, the evidence regarding mode of action includes direct

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service