Skip to Content
Merck
All Photos(1)

Key Documents

385891

Sigma-Aldrich

Benzenethionosulfonic acid sodium salt

technical grade, 85%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€214.00

€214.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€214.00

About This Item

Linear Formula:
C6H5S(=S)(=O)ONa
CAS Number:
Molecular Weight:
196.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€214.00


Check Cart for Availability

Request a Bulk Order

grade

technical grade

Quality Level

Assay

85%

mp

275 °C (dec.) (lit.)

functional group

sulfinic acid

SMILES string

[Na+].[O-]S(=O)(=S)c1ccccc1

InChI

1S/C6H6O2S2.Na/c7-10(8,9)6-4-2-1-3-5-6;/h1-5H,(H,7,8,9);/q;+1/p-1

InChI key

BZHOWMPPNDKQSQ-UHFFFAOYSA-M

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T R Chauncey et al.
The Journal of biological chemistry, 258(24), 15037-15045 (1983-12-25)
Thiosulfate reductase catalyzes the desulfuration of thiosulfonates while oxidizing GSH to GSSG. Kinetic studies of the enzyme-catalyzed reaction between GSH and benzenethiosulfonate have been carried out, and direct evidence for the occurrence of glutathione persulfide as an immediate product of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service