Skip to Content
Merck
All Photos(1)

Documents

385379

Sigma-Aldrich

Indium

rod, diam. 10 mm, 99.9998% trace metals basis

Synonym(s):

Indium element

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
In
CAS Number:
Molecular Weight:
114.82
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

vapor pressure

<0.01 mmHg ( 25 °C)

Assay

99.9998% trace metals basis

form

rod

resistivity

8.37 μΩ-cm

diam.

10 mm

mp

156.6 °C (lit.)

density

7.3 g/mL at 25 °C (lit.)

SMILES string

[In]

InChI

1S/In

InChI key

APFVFJFRJDLVQX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Quantity

26 g = 50 mm

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yongseok Kwon et al.
Organic letters, 15(4), 920-923 (2013-02-05)
This paper documents the first example of In(III)-catalyzed selective 6-exo-dig hydroarylation of o-propargylbiaryls and their subsequent double-bond migration to obtain functionalized phenanthrenes. Electron-rich biaryl substrates undergo hydroarylation more effectively, and the substrates with various types of substituents on the alkyne
R C Longo et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 25(8), 085506-085506 (2013-02-01)
Unlike graphene, a hexagonal InP sheet (HInPS) cannot be obtained by mechanical exfoliation from the native bulk InP, which crystallizes in the zinc blende structure under ambient conditions. However, by ab initio density functional theory calculations we found that a
Hwa Sub Oh et al.
Journal of nanoscience and nanotechnology, 13(1), 564-567 (2013-05-08)
We investigate Ga0.33In0.67P quantum dot structures appropriate for special lighting applications in terms of structural and optical behaviors. The Ga0.33In0.67P materials form from 2-dimentional to 3-dimensional dots as the nominal growth thickness increases from 0.5 nm to 6.0 nm, indicating
Juan Zhou et al.
Chemical communications (Cambridge, England), 49(22), 2237-2239 (2013-02-12)
A reduced graphene oxide (RGO)-ZnIn(2)S(4) nanosheet composite was successfully synthesized via an in situ controlled growth process. The as-obtained RGO-ZnIn(2)S(4) composite showed excellent visible light H(2) production activity in the absence of noble metal cocatalysts.
Ching-Hwa Ho et al.
ACS applied materials & interfaces, 5(6), 2269-2277 (2013-03-05)
The surface formation oxide assists of visible to ultraviolet photoelectric conversion in α-In2Se3 hexagonal microplates has been explored. Hexagonal α-In2Se3 microplates with the sizes of 10s to 100s of micrometers were synthesized and prepared by the chemical vapor transport method

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service