Skip to Content
Merck
All Photos(1)

Key Documents

384429

Sigma-Aldrich

tert-Butyldimethylsilyl chloride solution

1.0 M in methylene chloride

Synonym(s):

tert-Butylchlorodimethylsilane solution, tert-Butyldimethylchlorosilane, tert-Butyldimethylsilyl chloride, TBDMSCl solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CSi(CH3)2Cl
CAS Number:
Molecular Weight:
150.72
Beilstein:
505999
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

liquid

concentration

1.0 M in methylene chloride

density

0.87 g/mL at 20 °C (lit.)
1.225 g/mL at 25 °C

SMILES string

CC(C)(C)[Si](C)(C)Cl

InChI

1S/C6H15ClSi/c1-6(2,3)8(4,5)7/h1-5H3

InChI key

BCNZYOJHNLTNEZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Carc. 2 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

136.4 °F - closed cup

Flash Point(C)

58 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akshanth R Polepally et al.
Journal of clinical pharmacology, 55(10), 1101-1108 (2015-04-24)
A classic 2-period crossover bioavailability study was conducted to evaluate the relative and absolute bioavailability of immediate-release (IR) and extended-release (XR) lamotrigine formulations under steady-state conditions in elderly patients with epilepsy. On treatment days, each subject's morning dose (IR or
Some observations on the t-butyldimethylchlorosilane derivatization reaction.
K Bricknell et al.
Biochemical medicine, 23(1), 119-121 (1980-02-01)
Hiroaki Wakimoto et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 20(11), 2898-2909 (2014-04-10)
Isocitrate dehydrogenase (IDH) gene mutations occur in low-grade and high-grade gliomas. We sought to identify the genetic basis of malignant phenotype heterogeneity in IDH-mutant gliomas. We prospectively implanted tumor specimens from 20 consecutive IDH1-mutant glioma resections into mouse brains and
P Huttenloch et al.
Environmental science & technology, 35(21), 4260-4264 (2001-11-23)
The efficacy of the surface modification of natural diatomite and zeolite material by chlorosilanes is demonstrated. Chlorosilanes used were trimethylchlorosilane (TMSCI), tert-butyldimethylchlorosilane (TBDMSCI), dimethyloctadecylchlorosilane (DMODSCI), and diphenyldichlorosilane (DPDSCI) possessing different headgroups and chemical properties. Silanol groups of the diatomite and
C Tellis et al.
Journal of biochemistry, 119(4), 823-827 (1996-04-01)
1-O-Hexadecyl-2-acetyl-sn-glycerol, the immediate precursor of platelet- activating factor (PAF) in its de novo formation, was detected in the protozoon Tetrahymena pyriformis. It was purified from the total lipid extract by TLC, after successive developments in two different solvent systems. Characterization

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service