Skip to Content
Merck
All Photos(1)

Key Documents

383341

Sigma-Aldrich

Tungstosilicic acid hydrate

Tungstosilicic acid hydrate

≥99.9% trace metals basis

Synonym(s):

Silicotungstic acid

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€135.00

€135.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
€135.00

About This Item

Linear Formula:
H4[Si(W3O10)4] · xH2O
CAS Number:
Molecular Weight:
2878.17 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:
NACRES:
NA.22

€135.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥99.9% trace metals basis

form

powder, chunks or granules

composition

Degree of hydration, 25-26

SMILES string

O.O[Si](O)(O)O.O=[W]1(=O)O[W](=O)(=O)O[W](=O)(=O)O1.O=[W]2(=O)O[W](=O)(=O)O[W](=O)(=O)O2.O=[W]3(=O)O[W](=O)(=O)O[W](=O)(=O)O3.O=[W]4(=O)O[W](=O)(=O)O[W](=O)(=O)O4

InChI

1S/H4O4Si.H2O.36O.12W/c1-5(2,3)4;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/h1-4H;1H2;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;

InChI key

ACTPFSYIGLFGLY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Preparation of silicotungstate butylmethylimidazolium bromide ionic liquid hybrid material
  • Synthesis of SnII-substituted Keggin-type polyoxometalates as photocatalysts
  • Preparation of silicotungstic acid/organically modified silane proton-conducting membranes
  • Preparation of charge transfer compounds with Keggin anions
  • Preparation of catalyst for catalytic liquid-phase oxidation of xylene

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service