Skip to Content
Merck
All Photos(2)

Key Documents

383309

Sigma-Aldrich

Barium zirconate

powder, <10 μm

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€64.40

€64.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 G
€64.40

About This Item

Linear Formula:
BaZrO3
CAS Number:
Molecular Weight:
276.55
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

€64.40


Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

reaction suitability

reagent type: catalyst
core: zirconium

particle size

<10 μm

density

5.52 g/mL at 25 °C (lit.)

SMILES string

[Ba++].[O-][Zr]([O-])=O

InChI

1S/Ba.3O.Zr/q+2;;2*-1;

InChI key

DQBAOWPVHRWLJC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Barium zirconate (BaZrO₃) is a perovskite-structured ceramic known for its exceptional properties, including high thermal stability, low thermal expansion, and excellent ion conductivity. Its cubic crystal structure allows it to maintain stability at elevated temperatures, making it suitable for applications in thermal barrier coatings, proton-conducting fuelcells, and as a catalyst support in various chemical reactions. Additionally, BaZrO₃ exhibits low dielectric loss, making it attractive for electronic components and capacitors.

Application

  • Interface Diffusion and Compatibility of (Ba,La)FeO(3-δ) Perovskite Electrodes in Contact with Barium Zirconate and Ceria.: This study explores the interface diffusion and compatibility of perovskite electrodes in solid oxide fuel cells (SOFCs), demonstrating the stability of barium zirconate as an electrolyte material (Chiara et al., 2023).
  • Composite Electrolyte Used for Low Temperature SOFCs to Work at 390°C.: Researchers developed a composite electrolyte incorporating barium zirconate for use in low-temperature SOFCs, highlighting its potential in improving fuel cell performance (Liu et al., 2023).
  • In Situ Nanoscale Dynamics Imaging in a Proton-Conducting Solid Oxide for Protonic Ceramic Fuel Cells.: This article presents in situ imaging techniques to analyze the dynamics of proton conduction in barium zirconate, offering insights into its role in protonic ceramic fuel cells (Gorobtsov et al., 2022).
  • Proton Conductive BaZr(0.8-x) Ce(x) Y(0.2) O(3-δ): Influence of NiO Sintering Additive on Crystal Structure, Hydration Behavior, and Conduction Properties.: Investigating the impact of NiO additives, this study focuses on the proton conductivity and structural properties of yttrium-doped barium zirconate (Han et al., 2021).
  • Inkjet Printed Y-Substituted Barium Zirconate Layers as Electrolyte Membrane for Thin Film Electrochemical Devices.: The paper describes the application of inkjet printing techniques to create thin film electrolyte membranes using yttrium-doped barium zirconate for advanced electrochemical devices (Schneller et al., 2019).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Innovation in dental restorative materials is driven by the need for biocompatible and natural-appearing restoration alternatives. Conventional dental materials like amalgam and composite resins have inherent disadvantages.

A significant limiting factor for wearable electronics and wireless sensors is the finite amount of energy that can be stored in on-board batteries.

Among various ceramics, one-dimensional (1-D) piezoelectric ceramics have attracted significant scientific attention for use in energy harvesting.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service