Skip to Content
Merck
All Photos(1)

Key Documents

38263

Sigma-Aldrich

21-(Boc-amino)-4,7,10,13,16,19-hexaoxaheneicosanoic acid

purum, ≥97.0% (H-NMR)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H39NO10
CAS Number:
Molecular Weight:
453.52
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

purum

Assay

≥97.0% (H-NMR)

reaction suitability

reaction type: Pegylations
reagent type: cross-linking reagent

functional group

Boc
amine
carboxylic acid

storage temp.

2-8°C

SMILES string

OC(CCOCCOCCOCCOCCOCCOCCNC(OC(C)(C)C)=O)=O

InChI

1S/C20H39NO10/c1-20(2,3)31-19(24)21-5-7-26-9-11-28-13-15-30-17-16-29-14-12-27-10-8-25-6-4-18(22)23/h4-17H2,1-3H3,(H,21,24)(H,22,23)

InChI key

FTTYOIHYERRXQB-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Polyethylene glycol (PEG) reagents offer favorable characteristics for various applications, including biocompatibility and solubility.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service