Skip to Content
Merck
All Photos(3)

Key Documents

381225

Sigma-Aldrich

Sodium perchlorate hydrate

99.99% trace metals basis

Synonym(s):

Perchloric acid sodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaClO4 · xH2O
Molecular Weight:
122.44 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
crystalline
Pricing and availability is not currently available.

Quality Level

Assay

99.99% trace metals basis

form

crystalline

composition

Degree of hydration, ≤1

reaction suitability

reagent type: oxidant

impurities

≤150.0 ppm Trace Metal Analysis

SMILES string

[Na+].[O-]Cl(=O)(=O)=O

InChI

1S/ClHO4.Na.H2O/c2-1(3,4)5;;/h(H,2,3,4,5);;1H2/q;+1;/p-1

InChI key

IXGNPUSUVRTQGW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium perchlorate may be used to promote the hydrogen generation from reaction of tin with lactic acid by preventing the surface passivation of tin.[1]

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Ox. Sol. 1

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hydrogen generation by tin corrosion in lactic acid solution promoted by sodium perchlorate
Deyab MA.
Journal of Power Sources, 268, 765-770 (2014)
Laura A Hager et al.
Dalton transactions (Cambridge, England : 2003), 48(32), 12040-12049 (2019-07-12)
Quadruplex nucleic acids - DNA/RNA secondary structures formed in guanine rich sequences - proved to have key roles in the biology of cancers and, as such, in recent years they emerged as promising targets for small molecules. Many reports demonstrated
Katharina Ruthsatz et al.
Journal of applied toxicology : JAT, 38(11), 1416-1425 (2018-07-31)
Chemical, physical and biological environmental stressors may affect the endocrine system, such as the thyroid hormone (TH) axis in larval amphibians with consequences for energy partitioning among development, growth and metabolism. We studied the effects of two TH level affecting
Katharina Ruthsatz et al.
Journal of comparative physiology. B, Biochemical, systemic, and environmental physiology, 190(3), 297-315 (2020-03-08)
Environmental stress induced by natural and anthropogenic processes including climate change may threaten the productivity of species and persistence of populations. Ectotherms can potentially cope with stressful conditions such as extremes in temperature by exhibiting physiological plasticity. Amphibian larvae experiencing
Deniz Ucar et al.
Environmental technology, 37(2), 183-191 (2015-06-24)
Nitrate and perchlorate were identified as significant water contaminants all over the world. This study aims at evaluating the performances of the heterotrophic-autotrophic sequential denitrification process for reductive nitrate and perchlorate removal from drinking water. The reduced nitrate concentration in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service